Merck
  • Isolation of β-carotene, α-carotene and lutein from carrots by countercurrent chromatography with the solvent system modifier benzotrifluoride.
Isolation of β-carotene, α-carotene and lutein from carrots by countercurrent chromatography with the solvent system modifier benzotrifluoride.
Journal of chromatography. A (2015-03-03)
Michael Englert, Simon Hammann, Walter Vetter
ABSTRACT
A carotenoid purification method with dual-mode countercurrent chromatography (CCC) for β-carotene, α-carotene and lutein from a fresh carrot extract was developed. The fluorinated liquid benzotrifluoride (IUPAC name: (trifluoromethyl)benzene) was used as a novel modifier in the non-aqueous ternary solvent system n-hexane/benzotrifluoride/acetonitrile. The ternary phase diagram of the type I solvent system was used to select two-phase solvent mixtures which enabled an efficient preparative separation of α-carotene, β-carotene and lutein from concomitant pigments in crude carrot extract. By means of the modifier, high separation factors (α ≥ 1.2) were obtained, allowing baseline resolution between α-carotene and β-carotene due to specific chemical interactions such as π-π molecular interactions. After optimizing the injection step with a pseudo-ternary phase diagram, 51 mg of β-carotene, 32 mg of α-carotene and 4 mg of lutein could be isolated from 100.2mg crude carrot extract in a short time and with high purities of 95% and 99% by using dual-mode CCC, respectively. Temperatures > 22°C had a negative impact on the separation of α-carotene and β-carotene.
Sigma-Aldrich
(±)-α-Carotene, ≥95.0% (HPLC)