Merck
  • Potentiation of paclitaxel-induced apoptosis by the novel cyclin-dependent kinase inhibitor NU6140: a possible role for survivin down-regulation.
Potentiation of paclitaxel-induced apoptosis by the novel cyclin-dependent kinase inhibitor NU6140: a possible role for survivin down-regulation.
Molecular cancer therapeutics (2005-09-20)
Marzia Pennati, Allyson J Campbell, Maria Curto, Mara Binda, Yuzhu Cheng, Lan-Zeng Wang, Nicola Curtin, Bernard T Golding, Roger J Griffin, Ian R Hardcastle, Andrew Henderson, Nadia Zaffaroni, David R Newell
ABSTRACT
Cyclin-dependent kinases (CDK) play a crucial role in the control of the cell cycle. Aberrations in the control of cell cycle progression occur in the majority of human malignancies; hence, CDKs are promising targets for anticancer therapy. Here, we define the cellular effects of the novel CDK inhibitor NU6140, alone or in association with paclitaxel, with respect to inhibition of cell proliferation and cell cycle progression and induction of apoptosis in HeLa cervical carcinoma cells and in comparison with purvalanol A. Both CDK inhibitors induced a concentration-dependent cell cycle arrest at the G(2)-M phase and an increase in the apoptotic rate, with a concomitant down-regulation of the antiapoptotic protein survivin, a member of the inhibitors of apoptosis protein family. Notably, the addition of NU6140 to paclitaxel-treated cells resulted in markedly increased cytotoxic effect and apoptotic response in comparison with the paclitaxel-purvalanol A combination (86 +/- 11% and 37 +/- 8%, respectively). Similarly, the extent of caspase-9 and caspase-3 activation in paclitaxel-NU6140-treated cells was approximately 4-fold higher than after the paclitaxel-purvalanol A combination. Moreover, an almost complete abrogation of the expression of the active, Thr(34)-phosphorylated form of survivin was observed in cells exposed to the paclitaxel-NU6140 combination. A synergistic effect of the paclitaxel-NU6140 combination, as a consequence of survivin inhibition and increased activation of caspase-9 and caspase-3, was also observed in OAW42/e ovarian cancer line but not in the derived OAW42/Surv subline ectopically expressing survivin. Results from this study indicate that NU6140 significantly potentiates the apoptotic effect of paclitaxel, with inhibition of survivin expression/phosphorylation as the potential mechanism.
Sigma-Aldrich
NU6140, ≥98% (HPLC)