Merck
  • Levels of circulating myeloid subpopulations and of heme oxygenase-1 do not predict CD4(+) T cell recovery after the initiation of antiretroviral therapy for HIV disease.
Levels of circulating myeloid subpopulations and of heme oxygenase-1 do not predict CD4(+) T cell recovery after the initiation of antiretroviral therapy for HIV disease.
AIDS research and therapy (2014-09-03)
Lillian Seu, Gabriel M Ortiz, Trevor D Burt, Steven G Deeks, Jeffrey N Martin, Joseph M McCune
ABSTRACT
The level (or frequency) of circulating monocyte subpopulations such as classical (CD14(hi)CD16(-)) and non-classical (CD14(dim)CD16(+)) monocytes varies during the course of HIV disease progression and antiretroviral therapy (ART). We hypothesized that such variation and/or differences in the degree to which these cells expressed the immunoregulatory enzyme, heme oxygenase-1 (HO-1), would be associated with CD4(+) T cell recovery after the initiation of ART. This hypothesis was tested in a cross-sectional study of four groups of HIV-infected subjects, including those who were seronegative, untreated virologic controllers [detectable viral load (VL) of <1000 copies/mL], untreated virologic non-controllers [VL > 10,000 copies/mL], and ART-mediated virologic controllers [VL < 75 copies/mL]. A longitudinal analysis of ART-treated subjects was also performed along with regression analysis to determine which biomarkers were associated with and/or predictive of CD4(+) T cell recovery. Suppressive ART was associated with increased levels of classical monocyte subpopulations (CD14(hi)CD16(-)) and decreased levels of non-classical monocyte populations (CD14(dim)CD16(+)). Among peripheral blood mononuclear cells (PBMCs), HO-1 was found to be most highly up-regulated in CD14(+) monocytes after ex vivo stimulation. Neither the levels of monocyte subpopulations nor of HO-1 expression in CD14(+) monocytes were significantly associated with the degree of CD4(+) T cell recovery. Monocyte subpopulations and HO-1 gene expression were, however, restored to normal levels by suppressive ART. These results suggest that the level of circulating monocyte subpopulations and their expression of HO-1 have no evident relationship to CD4(+) T cell recovery after the initiation of ART.
Product Description
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Trypan Blue, Dye content 60 %, ≥80% (HPLC)
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, K ≤0.02%, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, reag. Ph. Eur., K ≤0.02%, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Sodium hydroxide solution, purum, ≥32%
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Sodium hydroxide, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium hydroxide, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%