Merck
  • Antioxidant and cytoprotective properties of partridgeberry polyphenols.
Antioxidant and cytoprotective properties of partridgeberry polyphenols.
Food chemistry (2014-08-31)
Khushwant S Bhullar, H P Vasantha Rupasinghe
ABSTRACT
Partridgeberry (Vaccinium vitis-idaea) is a polyphenol-rich berry of the Ericaceous family, grown in Newfoundland and Labrador province of Canada. The aims of this study were to identify extraction solvents for the maximum recovery of polyphenols, to establish fractionation technique for isolation of major sub-classes of polyphenols, and to evaluate antioxidant and cytoprotective properties of the partridgeberry polyphenol preparations. The acidified 70% acetone was identified as the ideal solvent for the maximum recovery of polyphenols from partridgeberry. Further, aqueous two-phase extraction, column chromatography and UPLC-MS/MS were employed to produce three partridgeberry polyphenol fractions, rich in either, anthocyanins, flavan-3-ols or flavonols. All the three PPF were potent antioxidants and displayed cytoprotective activity through the activation of nuclear factor erythroid 2-related factor 2 pathway, scavenging of reactive oxygen species, and inhibition of cellular death. The current study suggests that partridgeberry has numerous potential health implications in both prevention and amelioration of various diseases involving oxidative stress.
Sigma-Aldrich
Iron(III) chloride, anhydrous, powder, ≥99.99% trace metals basis
Sigma-Aldrich
Iron(III) chloride, sublimed grade, ≥99.9% trace metals basis
Sigma-Aldrich
Quercetin, ≥95% (HPLC), solid
Supelco
Quercetin, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Quercetin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Fluorescein, for fluorescence, free acid
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Millipore
TDA Reagent, suitable for microbiology
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
1,1-Diphenyl-2-picrylhydrazine, 97%
Sigma-Aldrich
s-Triazine, 97%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
2,2-Diphenyl-1-picrylhydrazyl
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Gallic acid, 97.5-102.5% (titration)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Iron(III) chloride solution, 0.2 M in 2-methyltetrahydrofuran
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Supelco
Gallic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%
Fluorescein, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%