Merck
In situ formation of H2O2 for P450 peroxygenases.
Bioorganic & medicinal chemistry (2014-07-06)
Caroline E Paul, Ekaterina Churakova, Elmer Maurits, Marco Girhard, Vlada B Urlacher, Frank Hollmann
ABSTRACT
An in situ H2O2 generation approach to promote P450 peroxygenases catalysis was developed through the use of the nicotinamide cofactor analogue 1-benzyl-1,4-dihydronicotinamide (BNAH) and flavin mononucleotide (FMN). Final productivity could be enhanced due to higher enzyme stability at low H2O2 concentrations. The H2O2 generation represented the rate-limiting step, however it could be easily controlled by varying both FMN and BNAH concentrations. Further characterization can result in an optimized ratio of FMN/BNAH/O2/biocatalyst enabling high reaction rates while minimizing H2O2-related inactivation of the enzyme.
Sigma-Aldrich
Lauric acid, natural, ≥98%, FCC, FG
Sigma-Aldrich
Myristic acid, natural, ≥98.5%, FG
Sigma-Aldrich
Diethyl ether, contains 1 ppm BHT as inhibitor, anhydrous, ≥99.7%
Sigma-Aldrich
Myristic acid, ≥98.0% (GC)
Supelco
Myristic acid, analytical standard
Supelco
Trizma® base, reference material for titrimetry, certified by BAM, >99.5%
Supelco
Diethyl ether, analytical standard
Sigma-Aldrich
Chlorotrimethylsilane, puriss., ≥99.0% (GC)
Sigma-Aldrich
Chlorotrimethylsilane, ≥98.0% (GC)
Sigma-Aldrich
Trizma® base, ≥99.0% (T)
Sigma-Aldrich
Trizma® base, BioUltra, for molecular biology, ≥99.8% (T)
Sigma-Aldrich
Lauric acid, ≥98%, FCC, FG
Sigma-Aldrich
Chlorotrimethylsilane, purified by redistillation, ≥99%
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ACS reagent, ≥99.8%
Sigma-Aldrich
Myristic acid, ≥95%, FCC, FG
Sigma-Aldrich
Diethyl ether, puriss. p.a., ACS reagent, ≥99.8% (GC)
Sigma-Aldrich
Myristic acid, Sigma Grade, ≥99%
Sigma-Aldrich
Trizma® base, ≥99.9% (titration), crystalline
Sigma-Aldrich
Trizma® base, BioXtra, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
Sigma-Aldrich
Trizma® base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Sigma-Aldrich
Tromethamine, meets USP testing specifications
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Trizma® base, puriss. p.a., ≥99.7% (T)
Sigma-Aldrich
Chlorotrimethylsilane, produced by Wacker Chemie AG, Burghausen, Germany, ≥99.0% (GC)
Supelco
Myristic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Trometamol, European Pharmacopoeia (EP) Reference Standard
Supelco
Chlorotrimethylsilane, for GC derivatization, LiChropur, ≥99.0% (GC)
Sigma-Aldrich
Trizma® base, anhydrous, free-flowing, Redi-Dri, ≥99.9%
Lauric acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Tromethamine, pharmaceutical secondary standard, certified reference material