Merck
  • Type II anti-CD20 mAb-induced lysosome mediated cell death is mediated through a ceramide-dependent pathway.
Type II anti-CD20 mAb-induced lysosome mediated cell death is mediated through a ceramide-dependent pathway.
Biochemical and biophysical research communications (2015-01-21)
Hua Ren, Chengwei Zhang, Liwei Su, Xin Bi, Chao Wang, Liang Wang, Bo Wu
ABSTRACT
In the past decade, monoclonal antibodies (mAbs) have revolutionized the treatment of non-Hodgkin lymphomas (NHLs). Although Fc-dependent mechanisms of mAb-mediated tumor clearance have been extensively studied, the ability of mAbs to directly evoke programmed cell death (PCD) and the underlying mechanisms involved remain unclear. It is well established that type II anti-CD20 mAb (Tositumomab) potently evoked PCD through a caspases-independent, lysosome-mediated process, which is related to homotypic adhesion (HA) in NHL cell lines. Herein, we reveal that the induction of ceramide generation by anti-CD20 mAbs directly correlates with their ability to induce PCD. The inhibition of ceramide abrogated Tositumomab-induced PCD indicating that ceramide is required for the execution of cell death. Further experimental results revealed that ceramide was generated downstream of mAb-induced HA and upstream of lysosome leakage. These findings provide further insights into a previously unrecognized role for ceramide generation in mediating PCD evoked by type II anti-CD20 mAbs in Burkitt's lymphoma cells. This newly characterized cell death pathway may potentially be exploited to eliminate malignant cells.
Product Description
USP
Valacyclovir Related Compound G, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains amylenes as stabilizer
Supelco
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
N-Acetyl-D-penicillamine, ≥99.0% (T)
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
(6-methoxy-1-naphthyl)acetic acid, AldrichCPR
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, amylene stabilized
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, puriss. p.a., reag. ISO, reag. Ph. Eur., 99.0-99.4% (GC)
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Valaciclovir impurity G, European Pharmacopoeia (EP) Reference Standard
Supelco
Chloroform, analytical standard
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
4-(Dimethylamino)pyridine, purum, ≥98.0% (NT)
Supelco
Residual Solvent - Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
4-(Dimethylamino)pyridine, ReagentPlus®, ≥99%