Merck
  • Membrane protein CNNM4-dependent Mg2+ efflux suppresses tumor progression.
Membrane protein CNNM4-dependent Mg2+ efflux suppresses tumor progression.
The Journal of clinical investigation (2014-10-28)
Yosuke Funato, Daisuke Yamazaki, Shin Mizukami, Lisa Du, Kazuya Kikuchi, Hiroaki Miki
ABSTRACT
Intracellular Mg(2+) levels are strictly regulated; however, the biological importance of intracellular Mg(2+) levels and the pathways that regulate them remain poorly understood. Here, we determined that intracellular Mg(2+) is important in regulating both energy metabolism and tumor progression. We determined that CNNM4, a membrane protein that stimulates Mg(2+) efflux, binds phosphatase of regenerating liver (PRL), which is frequently overexpressed in malignant human cancers. Biochemical analyses of cultured cells revealed that PRL prevents CNNM4-dependent Mg(2+) efflux and that regulation of intracellular Mg(2+) levels by PRL and CNNM4 is linked to energy metabolism and AMPK/mTOR signaling. Indeed, treatment with the clinically available mTOR inhibitor rapamycin suppressed the growth of cancer cells in which PRL was overexpressed. In ApcΔ(14/+) mice, which spontaneously form benign polyps in the intestine, deletion of Cnnm4 promoted malignant progression of intestinal polyps to adenocarcinomas. IHC analyses of tissues from patients with colon cancer demonstrated an inverse relationship between CNNM4 expression and colon cancer malignancy. Together, these results indicate that CNNM4-dependent Mg(2+) efflux suppresses tumor progression by regulating energy metabolism.
Sigma-Aldrich
Citrate Concentrated Solution, BioReagent, suitable for coagulation assays, 4 % (w/v)
Supelco
Sodium Citrate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Citric acid trisodium salt, ≥98% (GC), anhydrous
Sigma-Aldrich
Citrate Concentrated Solution, BioUltra, for molecular biology, 1 M in H2O
Millipore
ANTI-FLAG® antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Actin, α-Smooth Muscle - Cy3 antibody, Mouse monoclonal, clone 1A4, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Nitrogen, ≥99.998%
Sigma-Aldrich
Monoclonal Anti-β-Tubulin antibody produced in mouse, clone TUB 2.1, ascites fluid