Merck
  • Antioxidant and anticholinesterase activities of five wild mushroom species with total bioactive contents.
Antioxidant and anticholinesterase activities of five wild mushroom species with total bioactive contents.
Pharmaceutical biology (2015-02-28)
Gulsen Tel, Mehmet Ozturk, Mehmet E Duru, Aziz Turkoglu
ABSTRACT
Recently, mushrooms are interesting natural products to be investigated due to exhibiting various bioactivities. This study determines the antioxidant and anticholinesterase activities of various extracts of five wild mushroom species. In addition, the total bioactive contents, namely, ascorbic acid, β-carotene, and lycopene along with phenolic and flavonoid contents were also determined spectrophotometrically. Antioxidant activity was tested by using five complementary tests; namely, β-carotene-linoleic acid, DPPH(•) scavenging, ABTS(•+) scavenging, cupric-reducing antioxidant capacity (CUPRAC), and metal chelating assays. The in vitro anticholinesterase activity was tested against acetylcholinesterase (AChE) and butyrylcholinesterase (BChE) enzymes using the Ellman method. The spectrophotometric methods were used to determine the total phenolic, flavonoid, ascorbic acid, β-carotene, and lycopene contents. The current study has shown that ethyl acetate extracts of Ganoderma lucidum (Curtis) P. Karst (IC50: 1.55 ± 0.05 µg/mL) and Funalia trogii (Berk.) Bondartsev & Singer (IC50: 4.31 ± 0.18 µg/mL) exhibited good lipid peroxidation inhibitory activity. The DPPH, ABTS, and CUPRAC assays supported this activity. The ethyl acetate and methanol extracts of Funalia trogii and Ganoderma lucidum indicated good anticholinesterase activity. Ganoderma lucidum had rich phenolic and flavonoid contents, indicating 98.67 ± 0.32 mg PEs/g extract and 160.38 ± 1.25 mg QEs/g extract, respectively. The results demonstrate that some of the mushroom species tested herein could be used in food and pharmaceutical industries as natural antioxidants.
Sigma-Aldrich
Quercetin, ≥95% (HPLC), solid
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
3-tert-Butyl-4-hydroxyanisole, ≥98% (sum of isomers, GC), ≤10% 2-BHA basis (GC)
Supelco
Linoleic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, 99%
Supelco
Methanol, analytical standard
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
Pyrocatechol, purified by sublimation, ≥99.5%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ReagentPlus®, 99%
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
Neocuproine, ≥98%
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ≥98%, BioReagent, suitable for determination of sulfhydryl groups
Sigma-Aldrich
Pyrocatechol, ≥99%
Sigma-Aldrich
Linoleic acid, ≥99%
Sigma-Aldrich
Linoleic acid, liquid, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
Linoleic acid, technical, 58-74% (GC)
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
USP
Quercetin, United States Pharmacopeia (USP) Reference Standard