Merck
  • PADI4 acts as a coactivator of Tal1 by counteracting repressive histone arginine methylation.
PADI4 acts as a coactivator of Tal1 by counteracting repressive histone arginine methylation.
Nature communications (2014-05-31)
Stephan Kolodziej, Olga N Kuvardina, Thomas Oellerich, Julia Herglotz, Ingo Backert, Nicole Kohrs, Estel la Buscató, Sandra K Wittmann, Gabriela Salinas-Riester, Halvard Bonig, Michael Karas, Hubert Serve, Ewgenij Proschak, Jörn Lausen
ABSTRACT
The transcription factor Tal1 is a critical activator or repressor of gene expression in hematopoiesis and leukaemia. The mechanism by which Tal1 differentially influences transcription of distinct genes is not fully understood. Here we show that Tal1 interacts with the peptidylarginine deiminase IV (PADI4). We demonstrate that PADI4 can act as an epigenetic coactivator through influencing H3R2me2a. At the Tal1/PADI4 target gene IL6ST the repressive H3R2me2a mark triggered by PRMT6 is counteracted by PADI4, which augments the active H3K4me3 mark and thus increases IL6ST expression. In contrast, at the CTCF promoter PADI4 acts as a repressor. We propose that the influence of PADI4 on IL6ST transcription plays a role in the control of IL6ST expression during lineage differentiation of hematopoietic stem/progenitor cells. These results open the possibility to pharmacologically influence Tal1 in leukaemia.
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
SAFC
BIS-TRIS
SAFC
BIS-TRIS
Sigma-Aldrich
Anti-TAL-1 Antibody, clone BTL73, clone BTL73, Upstate®, from mouse
Sigma-Aldrich
Anti-CARM1 Antibody, from rabbit
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Iodoacetamide, BioUltra
Sigma-Aldrich
Iodoacetamide, Single use vial of 56 mg
Sigma-Aldrich
Iodoacetamide, ≥99% (NMR), crystalline
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Anti-dimethyl-Histone H3 (Arg2) Antibody, serum, Upstate®
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Iodoacetamide, Vetec, reagent grade, 98%
SAFC
Iodoacetamide