Merck
  • Is hyperuricemia a risk factor for arteriosclerosis? Uric acid and arteriosclerosis in apolipoprotein e-deficient mice.
Is hyperuricemia a risk factor for arteriosclerosis? Uric acid and arteriosclerosis in apolipoprotein e-deficient mice.
Biological & pharmaceutical bulletin (2014-12-03)
Hirokazu Wakuda, Shinya Uchida, Masahiko Ikeda, Masaki Tabuchi, Yasumitsu Akahoshi, Kazumasa Shinozuka, Shizuo Yamada
ABSTRACT
Although hyperlipidemia, high blood pressure, and diabetes increase the risk of arteriosclerosis, it is not clear whether hyperuricemia increases the risk of arteriosclerosis or not. We examined the effects of uric acid and curative drugs for hyperuricemia on atherosclerosis-susceptible C57BL/6J apolipoprotein E-deficient (apoE(-/-)) mice. Male apoE(-/-) mice (age: 6 weeks) were fed a normal diet (normal diet group) or a uric acid-enriched diet. Mice fed the uric acid-enriched diet were divided into three groups and administered a drinking vehicle (high uric acid diet group), allopurinol (20 mg·kg(-1)·d(-1)), or benzbromarone (20 mg·kg(-1)·d(-1)) for 10 weeks. Serum uric acid concentrations were higher in the high uric acid diet group than in the normal diet group, and concentrations in the allopurinol and benzbromarone groups were lower than in the high uric acid diet group. Serum total cholesterol and triglyceride levels were lower in the allopurinol group than in the high uric acid diet group. Oxidative stress was lower in the benzbromarone group than in the high uric acid diet group. Atherosclerotic lesion areas were smaller in the allopurinol and benzbromarone groups than in the high uric acid diet group. Thus, hyperuricemia may not be an independent risk factor for arteriosclerosis; however, the administration of allopurinol and benzbromarone prevented the development of atherosclerosis in apoE(-/-) mice fed a uric acid-enriched diet. The anti-atherosclerotic effect was in part due to lower total cholesterol and oxidative stress in the serum. Other possible mechanisms underlying this effect should be investigated.
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Supelco
Benzbromarone, analytical standard
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Sodium phosphate dibasic solution, BioUltra, 0.5 M in H2O
Benzbromarone, European Pharmacopoeia (EP) Reference Standard
Supelco
Phosphate Standard for IC, TraceCERT®, 1000 mg/L phosphate in water (nominal concentration)
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Sodium phosphate dibasic, 99.95% trace metals basis
Sigma-Aldrich
Sodium phosphate dibasic, purum p.a., anhydrous, ≥98.0% (T)
Sigma-Aldrich
Sodium phosphate dibasic, BioUltra, for molecular biology, ≥99.5% (T)
SAFC
Cholesterol, Plant-Derived, SyntheChol®
Sigma-Aldrich
Potassium hydride, 30 wt % dispersion in mineral oil
Sigma-Aldrich
Sodium phosphate dibasic, for molecular biology, ≥98.5% (titration)
Sigma-Aldrich
Sodium phosphate dibasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Allopurinol, xanthine oxidase inhibitor
Sigma-Aldrich
Sodium phosphate dibasic, BioXtra, ≥99.0%