Merck
  • Icariin may benefit the mesenchymal stem cells of patients with steroid-associated osteonecrosis by ABCB1-promoter demethylation: a preliminary study.
Icariin may benefit the mesenchymal stem cells of patients with steroid-associated osteonecrosis by ABCB1-promoter demethylation: a preliminary study.
Osteoporosis international : a journal established as result of cooperation between the European Foundation for Osteoporosis and the National Osteoporosis Foundation of the USA (2014-08-13)
Z-B Sun, J-W Wang, H Xiao, Q-S Zhang, W-S Kan, F-B Mo, S Hu, S-N Ye
ABSTRACT
In this study, we found out a previously undefined function of icariin which restored the dynamic balance between osteogenic and adipogenic differentiation of mesenchymal stem cells (MSCs) in patients with osteonecrosis of femoral head (ONFH) via ABCB1-promoter demethylation. These findings provided important information regarding potential implication of icariin targeting epigenetic changes for the treatment of steroid -associated ONFH. Here, we investigated whether icariin can also exert a beneficial role in the reactivation of MSCs in the patients with steroid-associated ONFH via ABCB1-promoter demethylation. Bone marrow was collected from the proximal femur in patients with steroid-associated ONFH (n = 20) and patients with new femoral neck fractures (n = 22), and then MSCs were isolated. We investigated cell viability, intracellular reactive oxygen species (ROS) level, mitochondrial membrane potential (MMP), P-glycoprotein (P-gp) activity, the transcript levels of ABCB1 and oxidative stress-related genes, methylation extent at CpG islands of ABCB1 promoter, and osteogenic and adipogenic differentiation ability of MSCs from the femoral neck fractures group and from the steroid-associated ONFH group treated with or without icariin. We observed that MSCs from the steroid-associated ONFH group showed reduced proliferation ability, elevated ROS level, depressed MMP, weakened osteogenesis, and enhanced adipogenesis while low P-gp activity, transcription level of ABCB1, and oxidative stress-related genes as well as aberrant CpG islands hypermethylation of ABCB1 were also noted in steroid-associated ONFH group. Treatment with icariin obviously induced de novo P-gp expression, decreased oxidative stress, and promoted osteogenesis. Icariin may be a potential drug targeting epigenetic changes for the treatment of steroid-associated ONFH.
Sigma-Aldrich
JC-1, powder or solid (Crystals)
Sigma-Aldrich
Rhodamine 123, BioReagent, for fluorescence, ≥85% (HPLC)
Sigma-Aldrich
Rhodamine 123, mitochondrial specific fluorescent dye
Sigma-Aldrich
Sodium bisulfite solution, purum, ~40%
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, Vetec, reagent grade, 98%