Merck
  • Nebulization of active pharmaceutical ingredients with the eFlow(®) rapid: impact of formulation variables on aerodynamic characteristics.
Nebulization of active pharmaceutical ingredients with the eFlow(®) rapid: impact of formulation variables on aerodynamic characteristics.
Journal of pharmaceutical sciences (2014-07-06)
Moritz Beck-Broichsitter, Nadine Prüfer, Nina Oesterheld, Werner Seeger, Thomas Schmehl
ABSTRACT
Nebulization of active pharmaceutical ingredient (API) solutions is a well-established means to achieve pulmonary drug deposition. The current study identified the impact of formulation variables on the aerosolization performance of the eFlow(®) rapid with special respect to optimized lung application. API formulations (including excipient-supplemented samples) were investigated for physicochemical properties, then nebulized using vibrating-mesh technology. The generated aerosol clouds were analyzed by laser diffraction. Aerosol deposition characteristics in the human respiratory tract were estimated using an algebraic model. Remarkable effects on aerosolization performance [i.e., mass median aerodynamic diameter (MMAD)] of API solutions were obtained when the sample conductivity (by API concentration and type, sodium chloride addition) and dynamic viscosity (by application of sucrose and poly(ethylene glycol) 200) were elevated. A similar influence was observed for a decline in surface tension (by ethanol addition). Thus, a defined adjustment of formulation parameters allowed for a decrease of the MMAD from ∼ 8.0 μm to values as small as ∼ 3.5 μm. Consequently, the pattern and efficiency of aerosol deposition in the human respiratory tract were improved. In conclusion, identification of physicochemical variables and their way of influencing vibrating-mesh nebulization has been provided to deliver a platform for tailoring aerosol characteristics and thus, advancing pulmonary therapy.
Product Description
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ethylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
SAFC
Sodium chloride solution, 5 M
USP
Ethylene glycol, United States Pharmacopeia (USP) Reference Standard
Salbutamol sulfate, European Pharmacopoeia (EP) Reference Standard
Ipratropium bromide, European Pharmacopoeia (EP) Reference Standard
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Supelco
Ethylene glycol, analytical standard
Sigma-Aldrich
Ethylene glycol, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Supelco
Albuterol sulfate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethylene glycol, ReagentPlus®, ≥99%
Sigma-Aldrich
Ethylene glycol, anhydrous, 99.8%
Sigma-Aldrich
Ethylene glycol, spectrophotometric grade, ≥99%