Merck
  • Profiling of drug binding proteins by monolithic affinity chromatography in combination with liquid chromatography-tandem mass spectrometry.
Profiling of drug binding proteins by monolithic affinity chromatography in combination with liquid chromatography-tandem mass spectrometry.
Journal of chromatography. A (2014-07-30)
Xuepei Zhang, Tongdan Wang, Hanzhi Zhang, Bing Han, Lishun Wang, Jingwu Kang
ABSTRACT
A new approach for proteome-wide profiling drug binding proteins by using monolithic capillary affinity chromatography in combination with HPLC-MS/MS is reported. Two immunosuppresive drugs, namely FK506 and cyclosporin A, were utilized as the experimental models for proof-of-concept. The monolithic capillary affinity columns were prepared through a single-step copolymerization of the drug derivatives with glycidyl methacrylate and ethylene dimethacrylate. The capillary chromatography with the affinity monolithic column facilitates the purification of the drug binding proteins from the cell lysate. By combining the capillary affinity column purification and the shot-gun proteomic analysis, totally 33 FK506- and 32 CsA-binding proteins including all the literature reported target proteins of these two drugs were identified. Among them, two proteins, namely voltage-dependent anion-selective channel protein 1 and serine/threonine-protein phosphatase PGAM5 were verified by using the recombinant proteins. The result supports that the monolithic capillary affinity chromatography is likely to become a valuable tool for profiling of binding proteins of small molecular drugs as well as bioactive compounds.
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Phosphoric acid-16O4 solution, 70 wt. % in D2O, 99.9 atom % 16O
Supelco
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9% (GC)
Sigma-Aldrich
Phosphoric acid, BioReagent, suitable for insect cell culture, 85%
Sigma-Aldrich
Sigma 7-9®, ≥99% (titration), crystalline
Sigma-Aldrich
Acrylamide solution, 40%, suitable for electrophoresis, sterile-filtered
Sigma-Aldrich
Phosphoric acid, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Phosphoric acid, crystalline, ≥99.999% trace metals basis
Sigma-Aldrich
Methylamine, ≥99.0%
Supelco
Acrylamide solution, 40% in H2O
Supelco
Toluene, analytical standard
Sigma-Aldrich
Phosphoric acid, BioUltra, ≥85% (T)
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Sigma-Aldrich
Phosphoric acid, ≥85 wt. % in H2O, ≥99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
FKBP1a/FKBP12 human, recombinant, expressed in E. coli, ≥95% (SDS-PAGE)
Sigma-Aldrich
Toluene, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Toluene, suitable for HPLC, 99.9%
Supelco
Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Toluene, Laboratory Reagent, ≥99.3%