Merck
  • A pharmacodynamic/pharmacokinetic study of ficlatuzumab in patients with advanced solid tumors and liver metastases.
A pharmacodynamic/pharmacokinetic study of ficlatuzumab in patients with advanced solid tumors and liver metastases.
Clinical cancer research : an official journal of the American Association for Cancer Research (2014-03-19)
Josep Tabernero, Maria Elena Elez, Maria Herranz, Isabel Rico, Ludmila Prudkin, Jordi Andreu, Jose Mateos, Maria Josep Carreras, May Han, James Gifford, Marc Credi, Wei Yin, Shefali Agarwal, Philip Komarnitsky, Jose Baselga
ABSTRACT
This study evaluated the safety, tolerability, pharmacodynamics, pharmacokinetics, and antitumor activity of ficlatuzumab, a humanized hepatocyte growth factor (HGF) inhibitory monoclonal antibody, as monotherapy in patients with advanced solid tumors and liver metastases. Patients with p-Met (phosphorylated c-Met)-positive tumors enrolled in three dose-escalation cohorts, receiving ficlatuzumab 2, 10, or 20 mg/kg once per 14-day cycle. Pharmacodynamic changes in liver tumor biopsies and serum, pharmacokinetics, safety, and clinical activity were assessed. No dose-limiting toxicities occurred in the 19 patients enrolled (n = 6, 2 mg/kg; n = 7, 10 mg/kg; n = 6, 20 mg/kg). The most frequent diagnosis was colorectal cancer (n = 15; 79%). The most common treatment-emergent adverse events were asthenia, peripheral edema, hepatic pain (32% each), and cough (26%). Laboratory abnormalities of decreased serum albumin were present in all patients. Ficlatuzumab at 20 mg/kg lowered median levels of tumor p-Met (-53%), p-ERK (-43%), p-Akt (-2%), and increased median HGF levels (+33%), at the last on-study time point relative to baseline. Mean serum HGF levels increased with ficlatuzumab dose and number of treatment cycles. Ficlatuzumab exhibited linear pharmacokinetics and long terminal half-life (7.4-10 days). Best overall response was stable disease in 28% of patients, including 1 patient with pancreatic cancer with stable disease >1 year. Ficlatuzumab exhibited good safety/tolerability and demonstrated ability to modulate the HGF/c-Met pathway and downstream signaling in the tumor in patients with advanced solid tumors. Safety, pharmacodynamic, and pharmacokinetic data for ficlatuzumab confirmed the recommended phase II dose of 20 mg/kg once per 14-day cycle.
Sigma-Aldrich
Human HGF ELISA Kit, for serum, plasma, cell culture supernatant and urine
Pricing and availability is not currently available.
Sigma-Aldrich
Human HGF ELISA Kit, for cell and tissue lysates
Pricing and availability is not currently available.
Sigma-Aldrich
Mouse HGF ELISA Kit, for cell and tissue lysates
Pricing and availability is not currently available.
Sigma-Aldrich
Mouse HGF ELISA Kit, for serum, plasma and cell culture supernatant
Pricing and availability is not currently available.
Sigma-Aldrich
Hepatocyte Growth Factor human, HGF, recombinant, expressed in HEK 293 cells, HumanKine®, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Hepatocyte Growth Factor human, HGF, recombinant, expressed in NSO cells, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Hepatocyte Growth Factor human, HGF, recombinant, expressed in Baculovirus infected High-5 cells, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
HGF mouse, recombinant, expressed in Hi-5 Insect cells, ≥95% (SDS-PAGE), ≥95% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Phosphotyrosine MET / Hepatocyte Growth Factor Receptor ELISA Kit
Pricing and availability is not currently available.
Sigma-Aldrich
Human MET / Hepatocyte Growth Factor Receptor ELISA Kit
Pricing and availability is not currently available.
Sigma-Aldrich
HGF human, recombinant, expressed in CHO cells, ≥97% (SDS-PAGE), ≥97% (HPLC)
Pricing and availability is not currently available.