Merck
  • Effect of nicorandil in patients with heart failure: a systematic review and meta-analysis.
Effect of nicorandil in patients with heart failure: a systematic review and meta-analysis.
Cardiovascular therapeutics (2014-10-17)
Fujie Zhao, Sandip Chaugai, Peng Chen, Yan Wang, Dao Wen Wang
ABSTRACT
It is unclear whether nicorandil, a metabolic therapeutic drug, can be applied clinically to therapy of heart failure (HF). This meta-analysis evaluated therapeutic effects of nicorandil on HF patients. We performed a systematic review and meta-analysis of published studies evaluating effect of nicorandil on HF patients. Studies were stratified according to controlled versus uncontrolled designs and analyzed using random-effects meta-analysis models. We identified a total of 20 studies with a total of 1222 patients. In five randomized controlled studies, nicorandil treatment resulted in reduction in all-cause mortality and hospitalization for cardiac causes (HR: 0.35, P < 0.001) and improved cardiac pump function (SMD: 0.31, P = 0.02). In 15 observational studies, nicorandil therapy increases cardiac pump function (SMD: 0.75, P < 0.001), improves NYHA functional class (WMD: -1.33, P < 0.001), decreases PCWP (WMD: -6.86 mm Hg, P < 0.001), and pulmonary arterial pressure (SMD: -0.84, P < 0.001). The use of nicorandil in HF patients exerts substantial beneficial effects, suggesting that it may be an additional therapeutic agent for HF.
Sigma-Aldrich
(+)-Biotin 4-nitrophenyl ester, 98%