Merck
  • Determination of nitroimidazole residues in aquaculture tissue using ultra high performance liquid chromatography coupled to tandem mass spectrometry.
Determination of nitroimidazole residues in aquaculture tissue using ultra high performance liquid chromatography coupled to tandem mass spectrometry.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2014-05-06)
Anna Gadaj, Valentina di Lullo, Helen Cantwell, Martin McCormack, Ambrose Furey, Martin Danaher
ABSTRACT
An UHPLC-MS/MS method was developed for the quantitative confirmatory analysis of residues of nitroimidazole drugs (dimetridazole, ipronidazole, metronidazole, ornidazole and ronidazole) and their corresponding hydroxy metabolites (HMMNI, ipronidazile-OH and metronidazole-OH) in aquaculture tissue. Samples were extracted by shaking in acetonitrile, water, MgSO4 and NaCl before being defatted with n-hexane pre-saturated with acetonitrile and concentrated under nitrogen. Nitroimidazole residues were determined by UHPLC-MS/MS operating in positive electrospray ionisation mode using a reversed phase BEH C18 column. The method was validated according to the EU Commission Decision 2002/657/EC guidelines. The following performance studies were carried out: specificity, selectivity, linearity, within laboratory repeatability (WLr)/reproducibility (WLR), accuracy, precision, decision limit (CCα), detection capability (CCβ), absolute recovery and stability. The analytical range of the method is 0.1-20 μg kg(-1). Accuracy and precision of the method, under within-laboratory reproducibility conditions, ranged from 83 to 105% and 2.3 to 14.0%, respectively. CCα were 0.07-1.0 μg kg(-1) depending on analyte and matrix. A total of 50 samples can be analysed in a single day using the assay. The method has been extensively evaluated through application to real test samples.
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Supelco
2-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Magnesium sulfate solution, BioUltra, for molecular biology
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Supelco
2-Propanol, analytical standard
Sigma-Aldrich
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
Supelco
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9% (GC)
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Hexane, analytical standard
Sigma-Aldrich
Magnesium sulfate solution, for molecular biology, 1.00 M±0.04 M
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Sigma-Aldrich
2-Propanol, for molecular biology, BioReagent, ≥99.5%
Supelco
Ipronidazole-OH, VETRANAL®, analytical standard
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
2-Propanol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
Hexane, puriss. p.a., ACS reagent, ≥99.0% (GC)
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Magnesium sulfate, puriss. p.a., drying agent, anhydrous, ≥98.0% (KT), powder (very fine)
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard