Merck
  • Fabrication and characterization of non-Brownian particle-based crystals.
Fabrication and characterization of non-Brownian particle-based crystals.
Langmuir : the ACS journal of surfaces and colloids (2014-07-02)
Melissa H Lash, Morgan V Fedorchak, Steven R Little, Joseph J McCarthy
ABSTRACT
Particle-based crystals have been explored in the literature for applications in molecular electronics, photonics, sensors, and drug delivery. However, much of the research on these crystals has been focused on particles of nano- and submicrometer dimensions (so-called colloidal crystals) with limited attention directed toward building blocks with dimensions ranging from tens to hundreds of micrometers. This can be attributed, in part, to the fact that the underlying thermal effects in these larger systems typically cannot naturally overcome kinetic barriers at the meso- and macroscales so that many of the methods used for nanoscale particle assembly cannot be directly applied to larger components, as they become kinetically arrested in nonequilibrium states. In this work, ultrasonic agitation is being explored as a means of allowing large, non-Brownian microparticles (18-750 μm) to overcome the kinetic barriers to packing in the creation of close-packed, highly ordered, crystalline structures. In addition, we study how the energy input affects bulk particle behavior and describe several new ways to characterize particle-based crystals made from microparticles.
Product Description
Supelco
Sulfuric acid, for the determination of nitrogen, ≥97.0%
Sigma-Aldrich
Sulfuric acid, 99.999%
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Sulfuric acid, puriss., meets analytical specification of Ph. Eur., BP, 95-97%
Sigma-Aldrich
Sulfuric acid, ACS reagent, 95.0-98.0%
Sigma-Aldrich
Sulfuric acid, puriss. p.a., for determination of Hg, ACS reagent, reag. ISO, reag. Ph. Eur., 95.0-97.0%
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
Sulfuric acid solution, puriss. p.a., ≥25% (T)
Supelco
Sulfuric acid concentrate, 0.1 M H2SO4 in water (0.2N), eluent concentrate for IC