Merck
  • Hypercaloric diets with increased meal frequency, but not meal size, increase intrahepatic triglycerides: a randomized controlled trial.
Hypercaloric diets with increased meal frequency, but not meal size, increase intrahepatic triglycerides: a randomized controlled trial.
Hepatology (Baltimore, Md.) (2014-03-29)
Karin E Koopman, Matthan W A Caan, Aart J Nederveen, Anouk Pels, Mariette T Ackermans, Eric Fliers, Susanne E la Fleur, Mireille J Serlie
ABSTRACT
American children consume up to 27% of calories from high-fat and high-sugar snacks. Both sugar and fat consumption have been implicated as a cause of hepatic steatosis and obesity but the effect of meal pattern is largely understudied. We hypothesized that a high meal frequency, compared to consuming large meals, is detrimental in the accumulation of intrahepatic and abdominal fat. To test this hypothesis, we randomized 36 lean, healthy men to a 40% hypercaloric diet for 6 weeks or a eucaloric control diet and measured intrahepatic triglyceride content (IHTG) using proton magnetic resonance spectroscopy ((1) H-MRS), abdominal fat using magnetic resonance imaging (MRI), and insulin sensitivity using a hyperinsulinemic euglycemic clamp with a glucose isotope tracer before and after the diet intervention. The caloric surplus consisted of fat and sugar (high-fat-high-sugar; HFHS) or sugar only (high-sugar; HS) and was consumed together with, or between, the three main meals, thereby increasing meal size or meal frequency. All hypercaloric diets similarly increased body mass index (BMI). Increasing meal frequency significantly increased IHTG (HFHS mean relative increase of 45%; P = 0.016 and HS mean relative increase of 110%; P = 0.047), whereas increasing meal size did not (2-way analysis of variance [ANOVA] size versus frequency P = 0.03). Abdominal fat increased in the HFHS-frequency group (+63.3 ± 42.8 mL; P = 0.004) and tended to increase in the HS-frequency group (+46.5 ± 50.7 mL; P = 0.08). Hepatic insulin sensitivity tended to decrease in the HFHS-frequency group while peripheral insulin sensitivity was not affected. A hypercaloric diet with high meal frequency increased IHTG and abdominal fat independent of caloric content and body weight gain, whereas increasing meal size did not. This study suggests that snacking, a common feature in the Western diet, independently contributes to hepatic steatosis and obesity. ( www.clinicaltrials.gov; nr.NCT01297738.)
Product Description
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
USP
Hydrocortisone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Hydrocortisone, British Pharmacopoeia (BP) Assay Standard
Hydrocortisone, European Pharmacopoeia (EP) Reference Standard
Hydrocortisone for peak identification, European Pharmacopoeia (EP) Reference Standard