Merck
  • Folate-targeted multifunctional amino acid-chitosan nanoparticles for improved cancer therapy.
Folate-targeted multifunctional amino acid-chitosan nanoparticles for improved cancer therapy.
Pharmaceutical research (2014-09-05)
Vítor M Gaspar, Elisabete C Costa, João A Queiroz, Chantal Pichon, Fani Sousa, Ilídio J Correia
ABSTRACT
Tumor targeting nanomaterials have potential for improving the efficiency of anti-tumoral therapeutics. However, the evaluation of their biological performance remains highly challenging. In this study we describe the synthesis of multifunctional nanoparticles decorated with folic acid-PEG and dual amino acid-modified chitosan (CM-PFA) complexed with DNA and their evaluation in organotypic 2D co-cultures of cancer-normal cells and also on 3D multicellular tumor spheroids models. The physicochemical characterization of CM-PFA multifunctional carriers was performed by FTIR, (1)H NMR and DLS. 2D co-culture models were established by using a 1:2 cancer-to-normal cell ratio. 3D organotypic tumor spheroids were assembled using micromolding technology for high throughput screening. Nanoparticle efficiency was evaluated by flow cytometry and confocal microscopy. The CM-PFA nanocarriers (126-176 nm) showed hemocompatibility and were internalized by target cells, achieving a 3.7 fold increase in gene expression. In vivo-mimicking 2D co-cultures confirmed a real affinity towards cancer cells and a negligible uptake in normal cells. The targeted nanoparticles penetrated into 3D spheroids to a higher extent than non-targeted nanocarriers. Also, CM-PFA-mediated delivery of p53 tumor suppressor promoted a decrease in tumor-spheroids volume. These findings corroborate the improved efficiency of this delivery system and demonstrate its potential for application in cancer therapy.
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
DL-Histidine, ≥99% (TLC)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
SAFC
HEPES
SAFC
HEPES
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Resorufin, 95%
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Arginine, European Pharmacopoeia (EP) Reference Standard
Histidine, European Pharmacopoeia (EP) Reference Standard