Merck
  • Synaptic upregulation and superadditive interaction of dopamine D2- and μ-opioid receptors after peripheral nerve injury.
Synaptic upregulation and superadditive interaction of dopamine D2- and μ-opioid receptors after peripheral nerve injury.
Pain (2014-09-23)
Zigor Aira, Teresa Barrenetxea, Itsaso Buesa, Juan Carlos Gómez-Esteban, Jon Jatsu Azkue
ABSTRACT
A sound strategy for improving the clinical efficacy of opioids involves exploiting positive interactions with drugs directed at other targets in pain pathways. The current study investigated the role of dopamine receptor D2 (D2R) in modulation of spinal dorsal horn excitability to noxious input, and interactions therein with μ-opioid receptor (MOR) in an animal model of neuropathic pain induced by spinal nerve ligation (SNL). C-fiber-evoked field potentials in the spinal dorsal horn were depressed concentration dependently by spinal superfusion with the D2R agonist quinpirole both in nerve-injured and sham-operated (control) rats. However, quinpirole-induced depression was significant at 10 μmol/L after SNL but only at 100 μmol/L in control rats. This quinpirole effect was completely abolished by MOR antagonist CTOP at subclinical concentration (1 μmol/L) in nerve-injured rats, but was unaltered in sham-operated rats. Nine days after SNL, D2R was upregulated to both presynaptic and postsynaptic locations in dorsal horn neurons, as revealed by double confocal immunofluorescence stainings for synaptophysin and PSD-95. In addition, D2R/MOR co-localization was increased after SNL. Co-administration of 1 μmol/L quinpirole, insufficient per se to alter evoked potentials, dramatically enhanced inhibition of evoked potentials by MOR agonist DAMGO, reducing the IC50 value of DAMGO by 2 orders of magnitude. The present data provide evidence of profound functional and subcellular changes in D2R-mediated modulation of noxious input after nerve injury, including positive interactions with spinal MOR. These results suggest D2R co-stimulation as a potential avenue to improve MOR analgesia in sustained pain states involving peripheral nerve injury.
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Calcium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Supelco
Calcium ion solution for ISE, 0.1 M Ca, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
CTOP, ≥97% (HPLC)
Sigma-Aldrich
Calcium chloride
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Calcium chloride, Vetec, reagent grade, 96%
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Supelco
Calcium standard for AAS, analytical standard, 1.000 g/L Ca+2 in hydrochloric acid, traceable to BAM
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate