Merck
  • Induced myogenic commitment of human chondrocytes via non-viral delivery of minicircle DNA.
Induced myogenic commitment of human chondrocytes via non-viral delivery of minicircle DNA.
Journal of controlled release : official journal of the Controlled Release Society (2015-01-03)
Jieun Hong, Eunjee A Lee, Eun-Seo Lee, Giyoung Jung, Hansaem Jeong, Hwajin Lee, Hyukjin Lee, Nathaniel S Hwang
ABSTRACT
Lineage conversion from one somatic cell type to another is an attractive approach for deriving specific therapeutic cell generation. In order to bypass inducing pluripotent stage, transdifferentiation/direct conversion technologies have been recently developed. We report the development of a direct conversion methodology in which cells are transdifferentiated through a plastic intermediate state induced by exposure to non-integrative minicircle DNA (MCDNA)-based reprogramming factors, followed by differentiation into myoblasts. In order to increase the MCDNA delivery efficiency, reprogramming factors were delivered into the chondrocytes via electroporation followed by poly (β-amino esters) (PBAE) transfection. We used this approach to convert human chondrocytes to myoblast, and with treatment of SB-431542, an inhibitor of the activin receptor-like kinase receptors, to enhance myogenic commitment. Differentiated cells exhibited expression of myogenic markers such as MyoD and Myog. This methodology for direct lineage conversion from chondrocytes to myoblast represents a novel non-viral Method to convert hard-to-transfect cells to other lineage.
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
L-Proline, Vetec, reagent grade, ≥99%
SAFC
HEPES
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, K ≤0.02%, ≥98.0% (T), pellets
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
L-Proline, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Proline, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
1,4-Butanediol diacrylate, technical grade, contains ~75 ppm hydroquinone as inhibitor
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
L-Proline, 99%, FCC, FG
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
SAFC
L-Proline
Sigma-Aldrich
L-(+)-Arabinose, ≥99% (GC)
SAFC
HEPES
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
L-Proline, from non-animal source, meets EP, USP testing specifications, suitable for cell culture
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%