Merck
  • Surface roughness and morphology of resin composites polished with two-step polishing systems.
Surface roughness and morphology of resin composites polished with two-step polishing systems.
Dental materials journal (2014-03-07)
Esra Can Say, Haktan Yurdagüven, Batu Can Yaman, Füsun Özer
ABSTRACT
The purpose of this study was to investigate surface roughness (Ra) and morphology of supra-nanofilled [Estelite Omega (EO), Estelite ∑ Quick (EQ)], micro-hybrid [Esthet.X HD (EHD), G-aenial (GAE)] and nano-hybrid [Clearfil Majesty Posterior (CMP), Charisma Diamond (CD), Beautifil II (BII)] composites polished with two-step polishing systems [Enhance/PoGo (EP); Venus Supra (VS)]. Composite discs, 30 of each type, were prepared. Each composite group was divided into three subgroups: Mylar (control), EP and VS. Ra was evaluated with profilometer. In each composite group, the control had the lowest Ra. With both polishing systems, EO and EQ had significantly the lowest Ra, whereas CMP and BII had the highest. Except for GAE, CD and BII, the differences in Ra between EP and VS in each composite group were significant, showing smoother surfaces for EP. Supra-nanofilled composites created smoother surfaces than nano-hybrids, and their performance was similar or slightly better than that of micro-hybrids.
Sigma-Aldrich
Aluminum oxide, nanopowder, <50 nm particle size (TEM)
Sigma-Aldrich
Aluminum oxide, nanowires, diam. × L 2-6 nm × 200-400 nm
Sigma-Aldrich
Aluminum oxide, nanoparticles, <50 nm particle size (DLS), 20 wt. % in isopropanol
Sigma-Aldrich
Aluminum oxide, nanopowder, 13 nm primary particle size (TEM), 99.8% trace metals basis
Sigma-Aldrich
Aluminum oxide, single crystal substrate, <0001>
Supelco
Aluminum oxide, for the determination of hydrocarbons
Sigma-Aldrich
Aluminum oxide, nanoparticles, 30-60 nm particle size (TEM), 20 wt. % in H2O
Sigma-Aldrich
Aluminum oxide, 99.997% trace metals basis
Sigma-Aldrich
Aluminum oxide, activated, basic, Brockmann I
Sigma-Aldrich
Aluminum oxide, activated, neutral, Brockmann I
Sigma-Aldrich
Aluminum oxide, activated, acidic, Brockmann I
Sigma-Aldrich
Aluminum oxide, powder, primarily α phase, ≤10 μm avg. part. size, 99.5% trace metals basis
Sigma-Aldrich
Aluminum oxide, Type WN-6, Neutral, Activity Grade Super I
Sigma-Aldrich
Aluminum oxide, pore size 58 Å, ~150 mesh
Sigma-Aldrich
Aluminum oxide, fused, powder, primarily α-phase, -325 mesh
Sigma-Aldrich
Aluminum oxide, powder, 99.99% trace metals basis
Sigma-Aldrich
Aluminum oxide, fused, powder, primarily α-phase, 100-200 mesh
Sigma-Aldrich
Aluminum oxide, Corundum, α-phase, -100 mesh
Sigma-Aldrich
Aluminum oxide, activated, acidic, Brockmann I, free-flowing, Redi-Dri
Sigma-Aldrich
Aluminum oxide, activated, neutral, Brockmann I, free-flowing, Redi-Dri
Sigma-Aldrich
Aluminum oxide, pellets, 3 mm
Sigma-Aldrich
Aluminum oxide, mesoporous, MSU-X (wormhole), average pore size 3.8 nm
Supelco
Aluminum oxide, activated, neutral, Brockmann Activity I