Merck
  • Three column intermittent simulated moving bed chromatography: 3. Cascade operation for center-cut separations.
Three column intermittent simulated moving bed chromatography: 3. Cascade operation for center-cut separations.
Journal of chromatography. A (2015-01-07)
Simon Jermann, Mattheus Meijssen, Marco Mazzotti
ABSTRACT
A general design methodology for chromatographic three fraction separation by application of the three column intermittent simulated moving bed (3C-ISMB) cascade is proposed and experimentally validated by studying the purification of an intermediately retained stereoisomer of nadolol, from an equimolar mixture of its four stereoisomers. The theoretical part shows that the 3C-ISMB cascade can be easily designed by applying Triangle Theory. Moreover, a re-scaling approach for the second stage is proposed so as to account for the fact that the feed flow rates to stage 2 are generally higher as compared to stage 1 due to dilution in the latter. Scaling the columns of the second stage accordingly enables to run both stages under optimal conditions with respect to switching time and step ratio, which is an important advantage as compared to integrated ternary processes. The experimental part starts with studying the linear adsorption behavior of nadolol in heptane/ethanol/DEA on Chiralpak AD for varying ratios of heptane and ethanol. Based on that, a solvent composition of Hept/EtOH/DEA 30/70/0.3 (v/v/v) is selected and the competitive multi-component Langmuir isotherm of the quaternary mixture is determined by frontal analysis. The resulting isotherm parameters are used to design several first stage experiments aiming at removal of the most retained component. The resulting ternary intermediate product is reprocessed in several second stage experiments studying various configurations. Finally, the dilution of the intermediate product with Hept/DEA yielding a solvent composition of Hept/EtOH/DEA 60/40/0.3 (v/v/v) is examined showing that the resulting increase in retention is beneficial for final product purities. Moreover, the reduction in viscosity compensates for the dilution as it enables higher flow rates. Dilution of the intermediate product is hence the best option, yielding highest overall cascade productivity (2.10gl(-1)h(-1)) and highest product purity (97.8%) requiring a specific solvent consumption of 12l/g of product.
Product Description
Supelco
Heptane, analytical standard
Sigma-Aldrich
Heptane, anhydrous, 99%
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Heptane, biotech. grade, ≥99%
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Heptane, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Heptane, ReagentPlus®, 99%
Sigma-Aldrich
Heptane, puriss. p.a., reag. Ph. Eur., ≥99% n-heptane basis (GC)
Sigma-Aldrich
Heptane, suitable for HPLC, ≥96%
Sigma-Aldrich
Heptane, HPLC Plus, for HPLC, GC, and residue analysis, 99%
Sigma-Aldrich
Heptane, suitable for HPLC, ≥99%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Supelco
Nadolol, analytical standard
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol, puriss. p.a., absolute, ≥99.8% (GC)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, HPLC/spectrophotometric grade
Sigma-Aldrich
Ethanol, ACS reagent, prima fine spirit, without additive, F15 o1
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 2% 2-butanone, A15 MEK1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Nadolol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%