Merck
  • Recombinant interleukin 6 with M cell-targeting moiety produced in Lactococcus lactis IL1403 as a potent mucosal adjuvant for peroral immunization.
Recombinant interleukin 6 with M cell-targeting moiety produced in Lactococcus lactis IL1403 as a potent mucosal adjuvant for peroral immunization.
Vaccine (2015-03-05)
Hui-Shan Li, Da-Chuan Piao, Tao Jiang, Jin-Duck Bok, Chong-Su Cho, Yoon-Seok Lee, Sang-Kee Kang, Yun-Jaie Choi
ABSTRACT
Development and application of safe and effective mucosal adjuvants are important to improve immunization efficiency in oral vaccine. Here, we report a novel mucosal adjuvant, IL-6-CKS9, a recombinant cytokine generated by conjugating an M cell-targeting peptide (CKS9) with c-terminus of the murine interleukin 6 (IL-6), which facilitated enhancement of mucosal immune response. Lactococcus lactis IL1403, a food-grade strain of lactic acid bacteria (LAB) which is widely used in dairy industry, was used as a host cell to express and secrete the IL-6-CKS9 for a mucosal vaccine adjuvant. The recombinant L. lactis IL1403 secreting IL-6-CKS9 was orally administered with a model antigen protein, M-BmpB (Brachyspira membrane protein B conjugated with CKS9), to BALB/c mice for mucosal immunization. ELISA analyses showed consistent enhancement tendencies in induction of anti-M-BmpB antibody levels both with mucosal (IgA) and systemic (IgG) immune responses in IL-6-CKS9-LAB treated group compared with other groups tested by conducting two separated mice immunization assays. In addition, we characterized that the oral administration of model protein antigen with live LAB producing IL-6-CKS9 could induce both Th1 and Th2 type immune responses by analysis of the specific anti-BmpB IgG1 and IgG2a isotypes in the sera and also investigated possible oral tolerance in our vaccine strategy. Collectively, our results showed successful production and secretion of recombinant murine IL-6 with M cell-targeting moiety (IL-6-CKS9) from L. lactis IL1403 and demonstrated the live recombinant LAB producing IL-6-CKS9 could have a potential to be used as an efficient adjuvant for peroral vaccination.
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥98.0% (NT)
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
3,3′,5,5′-Tetramethylbenzidine, standard for GC
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥99%
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Supelco
Acetone, analytical standard
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥98% (TLC)
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, tablet, 1 mg substrate per tablet
Sigma-Aldrich
2-Mercaptoethanol, for molecular biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Supelco
2-Mercaptoethanol, for HPLC derivatization, LiChropur, ≥99.0% (GC)
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Pyrvinium pamoate salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
DAPI, for nucleic acid staining