Merck
  • Lipophilic antioxidants prevent lipopolysaccharide-induced mitochondrial dysfunction through mitochondrial biogenesis improvement.
Lipophilic antioxidants prevent lipopolysaccharide-induced mitochondrial dysfunction through mitochondrial biogenesis improvement.
Pharmacological research (2014-12-03)
Pedro Bullón, Lourdes Román-Malo, Fabiola Marín-Aguilar, José Miguel Alvarez-Suarez, Francesca Giampieri, Maurizio Battino, Mario D Cordero
ABSTRACT
Oxidative stress is implicated in several infectious diseases. In this regard, lipopolysaccharide (LPS), an endotoxic component, induces mitochondrial dysfunction and oxidative stress in several pathological events such as periodontal disease or sepsis. In our experiments, LPS-treated fibroblasts provoked increased oxidative stress, mitochondrial dysfunction, reduced oxygen consumption and mitochondrial biogenesis. After comparing coenzyme Q10 (CoQ10) and N-acetylcysteine (NAC), we observed a more significant protection of CoQ10 than of NAC, which was comparable with other lipophilic and hydrophilic antioxidants such as vitamin E or BHA respectively. CoQ10 improved mitochondrial biogenesis by activating PGC-1α and TFAM. This lipophilic antioxidant protection was observed in mice after LPS injection. These results show that mitochondria-targeted lipophilic antioxidants could be a possible specific therapeutic strategy in pharmacology in the treatment of infectious diseases and their complications.
Product Description
Supelco
Rotenone, PESTANAL®, analytical standard
Sigma-Aldrich
N-Acetyl-L-cysteine, BioReagent, suitable for cell culture
Sigma-Aldrich
N-Acetyl-L-cysteine, Sigma Grade, ≥99% (TLC), powder
Sigma-Aldrich
L-(−)-Glucose, ≥99%
Sigma-Aldrich
Rotenone, ≥95%
Sigma-Aldrich
N-Acetyl-L-cysteine, BioXtra, ≥99% (TLC)
Sigma-Aldrich
2,4-Dinitrophenol, moistened with water, ≥98.0%
Supelco
2,4-Dinitrophenol, PESTANAL®, analytical standard
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)
Sigma-Aldrich
3-tert-Butyl-4-hydroxyanisole, ≥98% (sum of isomers, GC), ≤10% 2-BHA basis (GC)
Acetylcysteine, European Pharmacopoeia (EP) Reference Standard
USP
Acetylcysteine, United States Pharmacopeia (USP) Reference Standard
Supelco
3-tert-Butyl-4-hydroxyanisole, Pharmaceutical Secondary Standard; Certified Reference Material