Merck
  • Iodine quantification to distinguish clear cell from papillary renal cell carcinoma at dual-energy multidetector CT: a multireader diagnostic performance study.
Iodine quantification to distinguish clear cell from papillary renal cell carcinoma at dual-energy multidetector CT: a multireader diagnostic performance study.
Radiology (2014-08-28)
Achille Mileto, Daniele Marin, Marcela Alfaro-Cordoba, Juan Carlos Ramirez-Giraldo, Christian D Eusemann, Emanuele Scribano, Alfredo Blandino, Silvio Mazziotti, Giorgio Ascenti
ABSTRACT
To investigate whether dual-energy multidetector row computed tomographic (CT) imaging with iodine quantification is able to distinguish between clear cell and papillary renal cell carcinoma ( RCC renal cell carcinoma ) subtypes. In this retrospective, HIPAA-compliant, institutional review board-approved study, 88 patients (57 men, 31 women) with diagnosis of either clear cell or papillary RCC renal cell carcinoma at pathologic analysis, who underwent contrast material-enhanced dual-energy nephrographic phase study between December 2007 and June 2013, were included. Five readers, blinded to pathologic diagnosis, independently evaluated all cases by determining the lesion iodine concentration on color-coded iodine maps. The receiving operating characteristic curve analysis was adopted to estimate the optimal threshold for discriminating between clear cell and papillary RCC renal cell carcinoma , and results were validated by using a leave-one-out cross-validation. Interobserver agreement was assessed by using an intraclass correlation coefficient. The correlation between tumor iodine concentration and tumor grade was investigated. A tumor iodine concentration of 0.9 mg/mL represented the optimal threshold to discriminate between clear cell and papillary RCC renal cell carcinoma , and it yielded the following: sensitivity, 98.2% (987 of 1005 [95% confidence interval: 97.7%, 98.7%]); specificity, 86.3% (272 of 315 [95% confidence interval: 85.0%, 87.7%]); positive predictive value, 95.8% (987 of 1030 [95% confidence interval: 95.0%, 96.6%]); negative predictive value, 93.7% (272 of 290 [95% confidence interval: 92.8%, 94.7%]); overall accuracy of 95.3% (1259 of 1320 [95% confidence interval: 94.6%, 96.2%]), with an area under the curve of 0.923 (95% confidence interval: 0.913, 0.933). An excellent agreement was found among the five readers in measured tumor iodine concentration (intraclass correlation coefficient, 0.9990 [95% confidence interval: 0. 9987, 0.9993). A significant correlation was found between tumor iodine concentration and tumor grade for both clear cell (τ = 0.85; P < .001) and papillary RCC renal cell carcinoma (τ = 0.53; P < .001). Dual-energy multidetector CT with iodine quantification can be used to distinguish between clear cell and papillary RCC renal cell carcinoma , and it provides insights regarding the tumor grade.
Sigma-Aldrich
Iodine, 99.999% trace metals basis
Sigma-Aldrich
Iodine, anhydrous, beads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Iodine, ≥99.99% trace metals basis
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Sigma-Aldrich
Iodine, flakes, ReagentPlus®, ≥99%
Sigma-Aldrich
Iodine, puriss., ≥99.5% (RT), particles (round)
Sigma-Aldrich
Iodine, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.8-100.5%
Sigma-Aldrich
Iodine, ACS reagent, ≥99.8%, solid
Supelco
Iodine, ReagentPlus®, ≥99.8% (titration)
Sigma-Aldrich
Iodine, ReagentPlus®, 99.7% trace metals basis, beads, 1-3 mm