Merck
  • A cross-sectional analysis of candidate biomarkers of biological effect in smokers, never-smokers and ex-smokers.
A cross-sectional analysis of candidate biomarkers of biological effect in smokers, never-smokers and ex-smokers.
Biomarkers : biochemical indicators of exposure, response, and susceptibility to chemicals (2014-05-24)
Linsey E Haswell, Ermioni Papadopoulou, Nik Newland, Christopher J Shepperd, Frazer J Lowe
ABSTRACT
Biomarkers of biological effect (BOBE) have been proposed as potential tools to assess tobacco product use, toxicity and disease risk. To determine if candidate BOBE can distinguish between smokers, never-smokers and former smokers. Biomarker levels were compared from 143 smokers, 61 never-smokers and 61 ex-smokers. In total, 27 candidate biomarkers were assessed, 14 were significantly different between smokers and never-smokers (p < 0.01) and of these 14 biomarkers, 12 were able to distinguish between smokers and former smokers (p < 0.05), which indicates the potential for reversibility. A total of 12 of 27 BOBE are potentially useful tools for future product assessment.
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, Vetec, reagent grade, ≥98%
Sigma-Aldrich
Human sICAM1 ELISA Kit, for serum, plasma, cell culture supernatant and urine
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
Glutathione, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Mouse ICAM-1 ELISA Kit, for serum, plasma and cell culture supernatant