Merck
  • Polymer-based, flexible glutamate and lactate microsensors for in vivo applications.
Polymer-based, flexible glutamate and lactate microsensors for in vivo applications.
Biosensors & bioelectronics (2014-06-02)
Andreas Weltin, Jochen Kieninger, Barbara Enderle, Anne-Kathrin Gellner, Brita Fritsch, Gerald A Urban
ABSTRACT
We present a flexible microsensor, based on a polymer substrate, for multiparametric, electrochemical in vivo monitoring. The sensor strip with a microelectrode array at the tip was designed for insertion into tissue, for fast and localized online monitoring of physiological parameters. The microsystem fabrication on a wafer-level is based on a polyimide substrate and includes the patterning of platinum microelectrodes as well as epoxy and dry-film-resist insulation in a cost-effective thin-film and laminate process. A stable, electrodeposited silver/silver chloride reference electrode on-chip and a perm-selective membrane as an efficient interference rejection scheme are integrated on a wafer-level. Amperometric, electrochemical, enzyme-based biosensors for the neurotransmitter L-glutamate and the energy metabolite L-lactate have been developed. Hydrogel membranes or direct cross-linking as stable concepts for the enzyme immobilization are shown. Sensor performance including high selectivity, tailoring of sensitivity and long-term stability is discussed. For glutamate, a high sensitivity of 2.16 nAmm(-2) µM(-1) was found. For lactate, a variation in sensitivity between 2.6 and 32 nAmm(-2)mM(-1) was achieved by different membrane compositions. The in vivo application in an animal model is demonstrated by glutamate measurements in the brain of rats. Local glutamate alterations in the micromolar range and in nanoliter-range volumes can be detected and quantified with high reproducibility and temporal resolution. A novel, versatile platform for the integration of various electrochemical sensors on a small, flexible sensor strip for a variety of in vivo applications is presented.
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Supelco
Dopamine hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glutaraldehyde solution, technical, ~50% in H2O (5.6 M)
Dopamine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Ascorbic acid, 99%
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Dopamine hydrochloride
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Supelco
L-Ascorbic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material