Merck
  • Mineralization and bone regeneration using a bioactive elastin-like recombinamer membrane.
Mineralization and bone regeneration using a bioactive elastin-like recombinamer membrane.
Biomaterials (2014-07-06)
Esther Tejeda-Montes, Alexey Klymov, M Reza Nejadnik, Matilde Alonso, J Carlos Rodriguez-Cabello, X Frank Walboomers, Alvaro Mata
ABSTRACT
The search for alternative therapies to improve bone regeneration continues to be a major challenge for the medical community. Here we report on the enhanced mineralization, osteogenesis, and in vivo bone regeneration properties of a bioactive elastin-like recombinamer (ELR) membrane. Three bioactive ELRs exhibiting epitopes designed to promote mesenchymal stem cell adhesion (RGDS), mineralization (DDDEEKFLRRIGRFG), and both cell adhesion and mineralization were synthesized using standard recombinant protein techniques. The ELR materials were then used to fabricate membranes comprising either a smooth surface (Smooth) or channel microtopographies (Channels). Mineralization and osteoblastic differentiation of primary rat mesenchymal stem cells (rMSCs) were analyzed in both static and dynamic (uniaxial strain of 8% at 1 Hz frequency) conditions. Smooth mineralization membranes in static condition exhibited the highest quantity of calcium phosphate (Ca/P of 1.78) deposition with and without the presence of cells, the highest Young's modulus, and the highest production of alkaline phosphatase on day 10 in the presence of cells growing in non-osteogenic differentiation medium. These membranes were tested in a 5 mm-diameter critical-size rat calvarial defect model and analyzed for bone formation on day 36 after implantation. Animals treated with the mineralization membranes exhibited the highest bone volume within the defect as measured by micro-computed tomography and histology with no significant increase in inflammation. This study demonstrates the possibility of using bioactive ELR membranes for bone regeneration applications.
Product Description
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Supelco
L-Ascorbic acid, analytical standard
Supelco
N,N-Dimethylformamide, analytical standard
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Glutaraldehyde solution, technical, ~50% in H2O (5.6 M)
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
N,N-Dimethylformamide, suitable for HPLC, ≥99.9%
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
N,N-Dimethylformamide, for molecular biology, ≥99%
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Tetramethylsilane, electronic grade, ≥99.99% trace metals basis
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)