Merck
  • Modulation of BK channel activities by calcium-sensing receptor in rat bronchopulmonary sensory neurons.
Modulation of BK channel activities by calcium-sensing receptor in rat bronchopulmonary sensory neurons.
Respiratory physiology & neurobiology (2014-09-07)
Zhanna V Vysotskaya, Charles R Moss, Carolyn A Gilbert, Sabry A Gabriel, Qihai Gu
ABSTRACT
This study was carried out to investigate the expression of large-conductance Ca(2+)-activated potassium (BK) channels and to explore the possible modulation of BK channel activities by calcium-sensing receptors (CaSR) in rat bronchopulmonary sensory neurons. The expression of BK channels was demonstrated by immunohistochemistry and RT-PCR. Results from whole-cell patch-clamp recordings demonstrated that activation of CaSR with its agonist spermine or NPS R-568 showed a dual regulating effect on BK channel activities: it potentiated BK currents in cells exhibiting low baseline BK activity while slightly inhibited BK currents in cells with high baseline BK activity. Blocking CaSR with its antagonist NPS 2143 significantly inhibited BK currents. Our results further showed that the modulation of BK currents by CaSR activation or blockade was completely abolished when the intracellular Ca(2+) was chelated by BAPTA-AM. In summary, our data suggest that CaSR plays an integrative role in bronchopulmonary afferent signaling, at least partially through the regulation of BK channel activities.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, Vetec, reagent grade, 98%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Anti-KNG1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution