Merck
  • Gestational urinary bisphenol A and maternal and newborn thyroid hormone concentrations: the HOME Study.
Gestational urinary bisphenol A and maternal and newborn thyroid hormone concentrations: the HOME Study.
Environmental research (2015-03-22)
Megan E Romano, Glenys M Webster, Ann M Vuong, R Thomas Zoeller, Aimin Chen, Andrew N Hoofnagle, Antonia M Calafat, Margaret R Karagas, Kimberly Yolton, Bruce P Lanphear, Joseph M Braun
ABSTRACT
Bisphenol A (BPA), an endocrine disruptor used in consumer products, may perturb thyroid function. Prenatal BPA exposure may have sex-specific effects on thyroid hormones (THs). Our objectives were to investigate whether maternal urinary BPA concentrations during pregnancy were associated with THs in maternal or cord serum, and whether these associations differed by newborn sex or maternal iodine status. We measured urinary BPA concentrations at 16 and 26 weeks gestation among pregnant women in the HOME Study (2003-2006, Cincinnati, Ohio). Thyroid stimulating hormone (TSH) and free and total thyroxine (T4) and triiodothyronine (T3) were measured in maternal serum at 16 weeks (n=181) and cord serum at delivery (n=249). Associations between BPA concentrations and maternal or cord serum TH levels were estimated by multivariable linear regression. Mean maternal urinary BPA was not associated with cord THs in all newborns, but a 10-fold increase in mean BPA was associated with lower cord TSH in girls (percent change=-36.0%; 95% confidence interval (CI): -58.4, -1.7%), but not boys (7.8%; 95% CI: -28.5, 62.7%; p-for-effect modification=0.09). We observed no significant associations between 16-week BPA and THs in maternal or cord serum, but 26-week maternal BPA was inversely associated with TSH in girls (-42.9%; 95% CI: -59.9, -18.5%), but not boys (7.6%; 95% CI: -17.3, 40.2%; p-for-effect modification=0.005) at birth. The inverse BPA-TSH relation among girls was stronger, but less precise, among iodine deficient versus sufficient mothers. Prenatal BPA exposure may reduce TSH among newborn girls, particularly when exposure occurs later in gestation.
Sigma-Aldrich
Iodine, anhydrous, beads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Iodine, 99.999% trace metals basis
Sigma-Aldrich
Iodine, ≥99.99% trace metals basis
Sigma-Aldrich
Iodine, ACS reagent, ≥99.8%, solid
Supelco
Iodine, ReagentPlus®, ≥99.8% (titration)
Sigma-Aldrich
Iodine, puriss., ≥99.5% (RT), particles (round)
Sigma-Aldrich
Iodine, flakes, ReagentPlus®, ≥99%
Sigma-Aldrich
Iodine, ReagentPlus®, 99.7% trace metals basis, beads, 1-3 mm
Sigma-Aldrich
Iodine, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.8-100.5%