Merck
  • Effect of downscaling on the linearity range of a calibration curve in spectrofluorimetry.
Effect of downscaling on the linearity range of a calibration curve in spectrofluorimetry.
Analytical and bioanalytical chemistry (2014-05-13)
Radoslaw Kwapiszewski, Justyna Szczudlowska, Karina Kwapiszewska, Artur Dybko, Zbigniew Brzozka
ABSTRACT
Interest in the microfluidic environment, owing to its unique physical properties, is increasing in much innovative chemical, biological, or medicinal research. The possibility of exploiting and using new phenomena makes the microscale a powerful tool to improve currently used macroscopic methods and approaches. Previously, we reported that an increase in the surface area to volume ratio of a measuring cell could provide a wider linear range for fluorescein (Kwapiszewski et al., Anal. Bioanal. Chem. 403:151-155, 2012). Here, we present a broader study in this field to confirm the assumptions we presented before. We studied fluorophores with a large and a small Stokes shift using a standard cuvette and fabricated microfluidic detection cells having different surface area to volume ratios. We analyzed the effect of different configurations of the detection cell on the measured fluorescence signal. We also took into consideration the effect of concentration on the emission spectrum, and the effect of the surface area to volume ratio on the limit of linearity of the response of the selected fluorophores. We observed that downscaling, leading to an increase in the probability of collisions between molecules and cell walls with no energy transfer, results in an increase in the limit of linearity of the calibration curve of fluorophores. The results obtained suggest that microfluidic systems can be an alternative to the currently used approaches for widening the linearity of a calibration curve. Therefore, microsystems can be useful for studies of optically dense samples and samples that should not be diluted.
Product Description
Supelco
Glycine, analytical standard, for nitrogen determination according to Kjeldahl method
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Supelco
Glycine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Phosphate Standard for IC, TraceCERT®, 1000 mg/L phosphate in water (nominal concentration)
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycine, tested according to Ph. Eur.
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Glycine, 99%, FCC
SAFC
Glycine
Sigma-Aldrich
Eosin Y solution, 5 wt. % in H2O
Sigma-Aldrich
Eosin Y, 75% (HPLC)
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
4-Methylumbelliferone, ≥98%
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Eosin Y, Dye content ~99 %
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Quinine sulfate, meets USP testing specifications, monograph mol wt. 782.94 ((C20H24N2O2)2 . H2SO4 . 2H2O)
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Eosin Y disodium salt, certified by the Biological Stain Commission
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Eosin Y disodium salt, Dye content ≥85 %
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Sodium phosphate dibasic solution, BioUltra, 0.5 M in H2O