Merck
  • Practice patterns of phosphate binder use and their associations with mortality in chronic kidney disease.
Practice patterns of phosphate binder use and their associations with mortality in chronic kidney disease.
Clinical nephrology (2014-06-03)
Sankar D Navaneethan, Ankit Sakhuja, Susana Arrigain, John Sharp, Jesse D Schold, Joseph V Nally
ABSTRACT
Higher serum phosphorus is associated with an increased mortality among those with chronic kidney disease (CKD). We examined the practice patterns of phosphate binder use to lower serum phosphorus levels and their associations with mortality in the non-dialysisdependent CKD population. We examined the factors associated with the use of calcium and non-calcium phosphate binders in those with stage 3 and 4 CKD (eGFR 15 - 59 mL/min/1.73 m2) using logistic regression models. The associations between phosphate binder use and mortality were studied using propensity based analysis. Out of 57,928 patients with eGFR 15 - 59 mL/min/1.73 m2, 13,325 (23%) patients had serum phosphorus levels measured. 945 patients were prescribed phosphate binders, with 238 (25%) of them prescribed non-calcium-based phosphate binders and the rest calcium-based phosphate binders. Higher BMI, higher serum phosphorus, and higher serum calcium were associated with higher odds of being prescribed a non-calcium-based binder. Phosphate binder use was not significantly associated with mortality in either the entire cohort or the matched cohort in the analysis limited to those who were treated for at least 6 months. In the matched cohort, those who were treated for 1 year with a phosphate binder had a non-significant lower mortality rate (hazard ratio (HR): 0.85, 95% CI 0.66, 1.10). Phosphate binder use for 6 months and 1 year was not associated with reduced mortality in those with stage 3 and stage 4 CKD.
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), for GC derivatization, LiChropur
Supelco
Hydrogen chloride – ethanol solution, ~1.25 M HCl, for GC derivatization, LiChropur
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), for GC derivatization, LiChropur
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
USP
Calcium Carbonate (AS), United States Pharmacopeia (USP) Reference Standard
Supelco
Calcium Acetate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Hydrochloric acid, puriss., 24.5-26.0%
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Hydrochloric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., fuming, ≥37%, APHA: ≤10
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Calcium carbonate, 99.999% trace metals basis
Sigma-Aldrich
Calcium carbonate, BioUltra, precipitated, ≥99.0% (KT)
Supelco
Calcium carbonate, reference material for titrimetry, certified by BAM, ≥99.5%
Sigma-Aldrich
Calcium carbonate, ACS reagent, chelometric standard, 99.95-100.05% dry basis
Sigma-Aldrich
Calcium carbonate, tested according to Ph. Eur.
Sigma-Aldrich
Calcium carbonate, ≥99.995% trace metals basis
Sigma-Aldrich
Calcium carbonate, BioReagent, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Calcium carbonate, BioXtra, ≥99.0%
Supelco
Calcium carbonate, Pharmaceutical Secondary Standard; Certified Reference Material