Merck
  • miR-377 functions as a tumor suppressor in human clear cell renal cell carcinoma by targeting ETS1.
miR-377 functions as a tumor suppressor in human clear cell renal cell carcinoma by targeting ETS1.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie (2015-03-18)
Ruiyan Wang, Yanjie Ma, Dan Yu, Jiang Zhao, Peilong Ma
ABSTRACT
Clear cell renal cell carcinoma (ccRCC) is the most common form of neoplasm occurring in the adult kidney. Although significant advances have been made in treatment for ccRCC, a significant percent of patients have no benefit from currently available treatment option. MicroRNAs (miRNAs) have been reported to play central roles in regulating tumor progression, and are being explored as potentially more effective therapies and diagnostic tools for various cancers. The transcription factor E26 transformation specific-1 (ETS1) is believed to be intimately involved in tumor progression, and is frequently upregulated in tumors, including ccRCC. However, few studies have investigated the implications of ETS1 in ccRCC, and no studies have investigated the dysregulated mechanisms responsible for aberrant ETS1 expression in ccRCC. We used databases, clinical samples and target prediction algorithms to investigate aberrant miR-377 expression and potential targets in ccRCC. Our results indicate that miR-377 is downregulated in ccRCC, and that miR-377 can regulate the expression of ETS1. Further, using cell cycle analysis, MTT, luciferase and knockdown experiments, we found evidence to suggest that ETS1 is central in the development of the proliferative, metastatic and invasive phenotype of ccRCC cells. Furthermore, miR-377 was found to directly downregulate the expression of ETS1 and reduce the ability of ccRCC cells to proliferate, migrate and invade. As miR-377 was found to be differentially expressed in ccRCC, and in light of the apparent central role of ETS1 in tumor development, our results indicate that miR-377 could be useful for ccRCC diagnostics, prognostics and therapeutics.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium Fluoride Solution
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%