Merck
  • Species variation in the enantioselective metabolism of tegafur to 5-fluorouracil among rats, dogs and monkeys.
Species variation in the enantioselective metabolism of tegafur to 5-fluorouracil among rats, dogs and monkeys.
The Journal of pharmacy and pharmacology (2014-08-15)
Ikuo Yamamiya, Kunihiro Yoshisue, Yuji Ishii, Hideyuki Yamada, Ken-ichiro Yoshida
ABSTRACT
Tegafur (FT), a pro-drug of 5-fluorouracil (5-FU), is a racemate consisting of two enantiomers, R and S-FT. The aim of this study was to clarify interspecies variation in the enantioselective metabolism of FT. Plasma concentrations of FT enantiomers were determined in rats, dogs and monkeys following intravenous and oral dosing of the racemate (5 mg/kg). In addition, the enzymatic conversion of FT enantiomers to 5-FU was assayed using hepatic preparations. Metabolic clearance of R-FT was higher than that of S-FT in rats and monkeys, but S-FT was the preferential substrate for dogs. An inhibition study revealed that cytochrome P450 is primarily responsible for the enantioselective metabolism of FT in rats and dogs. In contrast, in monkeys, thymidine phosphorylase was a determinant of the enantioselectivity in FT metabolism. Although oral bioavailability was not enantioselective, in-vitro and in-vivo kinetic studies suggested that the enantioselectivity in the hepatic intrinsic clearance of FT directly influences the body clearance in all animal species examined. The interspecies variations were observed in the enantioselective pharmacokinetics of FT, and the in-vivo enantioselectivity could be extrapolated from the in-vitro metabolic activities.
Product Description
Supelco
Clotrimazole, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Chloramphenicol, VETRANAL®, analytical standard
Sigma-Aldrich
Magnesium chloride hexahydrate, 99.995% trace metals basis
Sigma-Aldrich
Magnesium chloride hexahydrate, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Quinidine, crystallized, ≥98.0% (dried material, NT)
Sigma-Aldrich
Quinine, suitable for fluorescence, anhydrous, ≥98.0% (dried material, NT)
Sigma-Aldrich
Magnesium chloride hexahydrate, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Quinine, 90%
Sigma-Aldrich
Chloramphenicol, BioReagent, suitable for plant cell culture
Sigma-Aldrich
Ketoconazole, 99.0-101.0% (EP, titration)
Sigma-Aldrich
Sulfaphenazole, ≥98%
Sigma-Aldrich
Clotrimazole
Sigma-Aldrich
Magnesium chloride hexahydrate, meets USP testing specifications
Sigma-Aldrich
Magnesium chloride hexahydrate, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Magnesium chloride hexahydrate, BioXtra, ≥99.0%
Sigma-Aldrich
Chloramphenicol, ≥98% (HPLC)
Sigma-Aldrich
Chloramphenicol, meets USP testing specifications
Sigma-Aldrich
Quinidine, anhydrous
Sigma-Aldrich
1-Aminobenzotriazole
Sigma-Aldrich
Sulfaphenazole, ≥98% (HPLC)
Supelco
Quinine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Clotrimazole for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Magnesium chloride hexahydrate, tested according to Ph. Eur.
Sigma-Aldrich
Magnesium chloride hexahydrate, puriss., meets analytical specification of Ph. Eur., BP, FCC, E511, 99-101%, ≤0.0001% Al
USP
Ketoconazole, United States Pharmacopeia (USP) Reference Standard
Clotrimazole, European Pharmacopoeia (EP) Reference Standard
Chloramphenicol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Magnesium chloride hexahydrate, ACS reagent, 99.0-102.0%
Supelco
Ketoconazole, Pharmaceutical Secondary Standard; Certified Reference Material
Ketoconazole, European Pharmacopoeia (EP) Reference Standard