Merck
  • Detection of 34 plasticizers and 25 flame retardants in indoor air from houses in Sapporo, Japan.
Detection of 34 plasticizers and 25 flame retardants in indoor air from houses in Sapporo, Japan.
The Science of the total environment (2014-04-29)
Shinji Takeuchi, Hiroyuki Kojima, Ikue Saito, Kazuo Jin, Satoshi Kobayashi, Toshiko Tanaka-Kagawa, Hideto Jinno
ABSTRACT
Various plasticizers and flame retardants are contained in building materials and furniture produced for indoor environments. However, some of these material inclusions have been reported to cause endocrine-disrupting and mucosa-irritating effects. Because of the local climate, buildings in Sapporo are better insulated against cold weather than those in many other areas in Japan. In this study, we measured 59 compounds, including plasticizers (phthalates, adipates, and others) and flame retardants (organo-phosphates and brominated compounds), from indoor air samples from six houses in Sapporo. These compounds were measured separately in the gas phase and the particle phase using a two-stage cartridge equipped with a quartz fiber filter (1 μm mesh) and C18 solid-phase extraction disk for sampling and analyzed by GC/MS and LC/MS/MS (for the detection of brominated flame retardants). Among the 59 compounds measured in this study, 34 compounds were detected from the indoor air of the six houses. The highest concentration among the 34 compounds found in a newly built house was 2,2,4-trimethyl-1,3-pentanediol monoisobutyrate (TXIB) at 20.8 μg/m(3). Di(2-ethyl-1-hexyl)terephthalate (DEHT), which has been used in recent years as an alternative to di(2-ethyl-1-hexyl)phthalate (DEHP), was found in all six houses, although at low concentrations ranging from 0.005 to 0.027 μg/m(3). To our knowledge, this is the first report of DEHT in indoor air in Japan. Among the compounds detected in this study, those with lower molecular weights tended to be captured in the C18 solid-phase extraction disk rather than in the quartz fiber filter. These results suggest that compounds with higher volatility exist preferentially in the gas phase, whereas compounds with lower volatility exist preferentially in the particulate phase in indoor air.
Sigma-Aldrich
Acetone, natural, ≥97%
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Supelco
Acetone, analytical standard
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Anthracene-d10, 98 atom % D