Merck
  • Design, synthesis, and biological evaluation of simplified side chain hybrids of the potent actin binding polyketides rhizopodin and bistramide.
Design, synthesis, and biological evaluation of simplified side chain hybrids of the potent actin binding polyketides rhizopodin and bistramide.
ChemMedChem (2015-02-03)
Daniel Herkommer, Sandra Dreisigacker, Galina Sergeev, Florenz Sasse, Holger Gohlke, Dirk Menche
ABSTRACT
The natural products rhizopodin and bistramide belong to an elite class of highly potent actin binding agents. They show powerful antiproliferative activities against a range of tumor cell lines, with IC50 values in the low-nanomolar range. At the molecular level they disrupt the actin cytoskeleton by binding specifically to a few critical sites of G-actin, resulting in actin filament stabilization. The important biological properties of rhizopodin and bistramide, coupled with their unique and intriguing molecular architectures, render them attractive compounds for further development. However, this is severely hampered by the structural complexity of these metabolites. We initiated an interdisciplinary approach at the interface between molecular modeling, organic synthesis, and chemical biology to support further biological applications. We also wanted to expand structure-activity relationship studies with the goal of accessing simplified analogues with potent biological properties. We report computational analyses of actin-inhibitor interactions involving molecular docking, validated on known actin binding ligands, that show a close match between the crystal and modeled structures. Based on these results, the ligand shape was simplified, and more readily accessible rhizopodin-bistramide mimetics were designed. A flexible and modular strategy was applied for the synthesis of these compounds, enabling diverse access to dramatically simplified rhizopodin-bistramide hybrids. This novel analogue class was analyzed for its antiproliferative and actin binding properties.
Product Description
Supelco
Water, for HPCE, for luminescence, suitable for UV/Vis spectroscopy
Sigma-Aldrich
Water, tested according to Ph. Eur.
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Supelco
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
Supelco
Water, for TOC analysis
Supelco
Water, suitable for ion chromatography
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Supelco
Water, ACS reagent, for ultratrace analysis
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Millipore
Urea solution, suitable for microbiology, 40% in H2O
Sigma-Aldrich
Water, BioPerformance Certified
Sigma-Aldrich
Water, ACS reagent
Sigma-Aldrich
Water, Deionized
Sigma-Aldrich
Water, suitable for HPLC
Sigma-Aldrich
Water, HPLC Plus
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O