Merck
  • Determination and separation of bisphenol A, phthalate metabolites and structural isomers of parabens in human urine with conventional high-pressure liquid chromatography combined with electrospray ionisation tandem mass spectrometry.
Determination and separation of bisphenol A, phthalate metabolites and structural isomers of parabens in human urine with conventional high-pressure liquid chromatography combined with electrospray ionisation tandem mass spectrometry.
Analytical and bioanalytical chemistry (2015-02-04)
Antonis Myridakis, Eirini Balaska, Christina Gkaitatzi, Antonis Kouvarakis, Euripides G Stephanou
ABSTRACT
Phthalates, bisphenol A (BPA) and parabens (PBs), organic chemicals widely used in everyday products, are considered to be endocrine disruptors. We propose a liquid chromatography-tandem mass spectrometry (LC-MS/MS) method for the determination of seven phthalate metabolites, six PBs and BPA in human urine. All three categories of the above endocrine disruptors were simultaneously extracted from 1 mL of human urine using solid phase extraction. In addition, with a conventional reversed phase LC column, we achieved for the first time the separation of three pairs of structural isomers, namely iso-/n-butyl paraben, propyl paraben and monobutyl phthalate. LC-MS/MS was operated and tested in both electrospray ionisation (ESI) and atmospheric pressure chemical ionisation (APCI). ESI was selected for the analysis due to its superior stability and repeatability. The method limit of detection (mLOD), achieved for a single set of high-performance LC conditions, ranged from 0.01 to 0.84 ng/mL for phthalate metabolites, from 0.06 to 0.24 ng/mL for PBs and was 2.01 ng/mL for BPA. Derivatisation of BPA with dansyl chloride lowered its mLOD to 0.007 ng/mL. Blank contamination was non-detectable. The present method was successfully applied for the analysis of the above-mentioned compounds in 80 male human urine samples.
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Millipore
Bifido Selective Supplement B, suitable for microbiology
Supelco
Butylparaben, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
Sigma-Aldrich
Phosphoric acid-16O4 solution, 70 wt. % in D2O, 99.9 atom % 16O
Sigma-Aldrich
Sodium phosphate monobasic solution, BioUltra, 5 M in H2O
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Phosphoric acid, ≥85 wt. % in H2O, ≥99.999% trace metals basis
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Sigma-Aldrich
Sodium phosphate monobasic, purum p.a., anhydrous, ≥99.0% (T)
Sigma-Aldrich
Ammonium hydroxide solution, BioUltra, ~1 M NH3 in H2O (T)
Sigma-Aldrich
Butyl 4-hydroxybenzoate, ≥99%
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Methyl 4-hydroxybenzoate, ≥99%, FCC
Sigma-Aldrich
Phosphoric acid, BioUltra, ≥85% (T)
Sigma-Aldrich
Propyl 4-hydroxybenzoate, tested according to Ph. Eur.
Sigma-Aldrich
Methyl 4-hydroxybenzoate, tested according to Ph. Eur.
Supelco
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9% (GC)
Sigma-Aldrich
Phosphoric acid, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Butyl 4-hydroxybenzoate, ≥99.0% (GC)
Sigma-Aldrich
Sodium phosphate monobasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Phosphoric acid, BioReagent, suitable for insect cell culture, 85%
Sigma-Aldrich
Methyl 4-hydroxybenzoate, analytical standard