Merck
  • Degradation pathways of low-ethoxylated nonylphenols by isolated bacteria using an improved method.
Degradation pathways of low-ethoxylated nonylphenols by isolated bacteria using an improved method.
Environmental science and pollution research international (2013-08-15)
Yu Zhang, Xin Gu, Jing Zhang, Min Yang
ABSTRACT
Nonylphenol ethoxylates (NPEOs) with low ethoxylation degree (NPav₂EO; containing two ethoxy units on average) and estrogenic properties are the intermediate products of nonionic surfactant NPEOs. To better understand the environmental fate of low-ethoxylated NPEOs, phylogenetically diverse low-ethoxylated NPEO-degrading bacteria were isolated from activated sludge using gellan gum as the gelling reagent. Four isolates belonging to four genera, i.e., Pseudomonas sp. NP522b in γ-Proteobacteria, Variovorax sp. NP427b and Ralstonia sp. NP47a in β-Proteobacteria, and Sphingomonas sp. NP42a in α-Proteobacteria were acquired. Ralstonia sp. NP47a or Sphingomonas sp. NP42a, have not been reported for the degradation of low-ethoxylated NPEOs previously. The biotransformation pathways of these isolates were investigated. The first three strains (NP522b, NP427b, and NP47a) exhibited high NPav₂EO oxidation ability by oxidizing the polyethoxy (EO) chain to form low-ethoxylated nonylphenoxy carboxylates, and then further oxidizing the alkyl chain to form carboxyalkylphenol polyethoxycarboxylates. Furthermore, Sphingomonas sp. NP42a degraded NPav2EO through a nonoxidative pathway with nonylphenol monoethoxylate as the dominant product.
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Chlorotrimethylsilane, puriss., ≥99.0% (GC)
Supelco
N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane, for GC derivatization, LiChropur, contains 10% TMCS, 98% (excluding TMCS)
Supelco
N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane, for GC derivatization, LiChropur, contains 1% TMCS, 99% (excluding TMCS)
Supelco
Methanol, analytical standard
Sigma-Aldrich
Ammonium acetate solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Ammonium acetate, BioUltra, for molecular biology, ≥99.0%
Sigma-Aldrich
Chlorotrimethylsilane, produced by Wacker Chemie AG, Burghausen, Germany, ≥99.0% (GC)
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Supelco
N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane, with 1% trimethylchlorosilane, for GC derivatization, LiChropur
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
Sigma-Aldrich
Chlorotrimethylsilane, ≥98.0% (GC)
Sigma-Aldrich
Chlorotrimethylsilane, purified by redistillation, ≥99%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Supelco
Chlorotrimethylsilane, for GC derivatization, LiChropur, ≥99.0% (GC)
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%