Merck
  • Long-term follow-up of successful hepatitis C virus therapy: waning immune responses and disappearance of liver disease are consistent with cure.
Long-term follow-up of successful hepatitis C virus therapy: waning immune responses and disappearance of liver disease are consistent with cure.
Alimentary pharmacology & therapeutics (2015-01-30)
M Hedenstierna, O Weiland, A Brass, D Bankwitz, P Behrendt, I Uhnoo, S Aleman, K Cardell, A Fryden, G Norkrans, A Eilard, H Glaumann, T Pietschmann, M Sällberg, E D Brenndörfer
ABSTRACT
A sustained viral response (SVR) after interferon-based therapy of chronic hepatitis C virus (HCV) infection is regarded to represent a cure. Previous studies have used different markers to clarify whether an SVR truly represents a cure, but no study has combined a clinical work-up with highly sensitive HCV RNA detection, and the determination of immune responses. To determine clinical, histological, virological and immunological markers 5-20 years after SVR. In 54 patients, liver biochemistry, histology and elastography were evaluated. Liver biopsies, plasma and peripheral blood mononuclear cells (PBMCs) were tested for minute amounts of HCV RNA. HCV-specific T-cell responses were monitored by ELISpot and pentamer staining, and humoral responses by measuring HCV nonstructural (NS)3-specific antibodies and virus neutralisation. Liver disease regressed significantly in all patients, and 51 were HCV RNA-negative in all tissues tested. There was an inverse association between liver disease, HCV-specific T-cell responses and HCV antibody levels with time from SVR, supporting that the virus had been cleared. The three patients, who all lacked signs of liver disease, had HCV RNA in PBMCs 5-9 years after SVR. All three had HCV-specific T cells and NS3 antibodies, but no cross-neutralising antibodies. Our combined data confirm that a SVR corresponds to a long-term clinical cure. The waning immune responses support the disappearance of the antigenic stimulus. Transient HCV RNA traces may be detected in some patients up to 9 years after SVR, but no marker associates this with an increased risk for liver disease.
Sigma-Aldrich
Sodium carbonate-12C, 99.9 atom % 12C
Supelco
Sodium carbonate concentrate, Na2CO3 72 mM in water, IC eluent concentrate (20x) for Metrosep A Supp 7
Supelco
Sodium carbonate concentrate, 0.1 M Na2CO3 in water, eluent concentrate for IC
Sigma-Aldrich
Phosphatase substrate, powder
Sigma-Aldrich
Phosphatase substrate, 40 mg tablets
Sigma-Aldrich
Phosphatase substrate, 40 mg capsules
Sigma-Aldrich
Phosphatase substrate, 100 mg capsules
Sigma-Aldrich
Phosphatase substrate, 5 mg tablets
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, tablet
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, tablet
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, tablet
Sigma-Aldrich
Sodium carbonate, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
Supelco
Sodium carbonate, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium carbonate, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, suitable for enzyme immunoassay, ≥99.0% (enzymatic)
Sigma-Aldrich
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent (primary standard), 99.95-100.05% dry basis
Sigma-Aldrich
Sodium carbonate, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium carbonate, anhydrous, powder or granules, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium carbonate, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, E500, anhydrous, 99.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium carbonate, ACS reagent, anhydrous, ≥99.5%, powder or granules
Sigma-Aldrich
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium carbonate, ACS reagent (primary standard), anhydrous, 99.95-100.05% dry basis
Sigma-Aldrich
Sodium carbonate, powder, ≥99.5%, ACS reagent
Sigma-Aldrich
Phosphatase substrate, Suitable for manufacturing of diagnostic kits and reagents