Merck
  • Comparison of pre-treatment and post-treatment use of selenium in retinal ischemia reperfusion injury.
Comparison of pre-treatment and post-treatment use of selenium in retinal ischemia reperfusion injury.
International journal of ophthalmology (2015-05-06)
Alper Yazici, Hasan Aksit, Esin Sogutlu Sari, Arzu Yay, Haydar Ali Erken, Dilek Aksit, Harun Cakmak, Kamil Seyrek, Sitki Samet Ermis
ABSTRACT
To investigate the effects of selenium in rat retinal ischemia reperfusion (IR) model and compare pre-treatment and post-treatment use. Selenium pre-treatment group (n=8) was treated with intraperitoneal (i.p.) selenium 0.5 mg/kg for 7d and terminated 24h after the IR injury. Selenium post-treatment group (n=8) was treated with i.p. selenium 0.5 mg/kg for 7d after the IR injury with termination at the end of the 7d period. Sham group (n=8) received i.p. saline injections identical to the selenium volume for 7d with termination 24h after the IR injury. Control group (n=8) received no intervention. Main outcome measures were retina superoxide dismutase (SOD), glutathione (GSH), total antioxidant status (TAS), malondialdehyde (MDA), DNA fragmentation levels, and immunohistological apoptosis evaluation. Compared to the Sham group, selenium pre-treatment had a statistical difference in all parameters except SOD. Post-treatment selenium also resulted in statistical differences in all parameters except the MDA levels. When comparing selenium groups, the pre-treatment selenium group had a statistically higher success in reduction of markers of cell damage such as MDA and DNA fragmentation. In contrast, the post-selenium treatment group had resulted in statistically higher levels of GSH. Histologically both selenium groups succeeded to limit retinal thickening and apoptosis. Pre-treatment use was statistically more successful in decreasing apoptosis in ganglion cell layer compared to post-treatment use. Selenium was successful in retinal protection in IR injuries. Pre-treatment efficacy was superior in terms of prevention of tissue damage and apoptosis.
Product Description
Glutathione, European Pharmacopoeia (EP) Reference Standard
Selenium, foil, 25x25mm, thickness 3mm, 99.95%
Selenium, pellets, < 5mm, ≥99.999%
Sigma-Aldrich
Sodium selenite, 99%
Sigma-Aldrich
Selenium, powder, −100 mesh, ≥99.5% trace metals basis
Sigma-Aldrich
Selenium, pellets, <5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Selenium, pellets, <5 mm particle size, ≥99.999% trace metals basis
Sigma-Aldrich
Sodium selenite, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Sodium selenite, γ-irradiated, lyophilized powder, BioXtra, suitable for cell culture
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium selenite, anhydrous, ≥90.0% (RT)
Sigma-Aldrich
Selenium, powder, −100 mesh, 99.99% trace metals basis
Sigma-Aldrich
L-Glutathione reduced, Vetec, reagent grade, ≥98%
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%