Merck
  • Differential modulation by Akkermansia muciniphila and Faecalibacterium prausnitzii of host peripheral lipid metabolism and histone acetylation in mouse gut organoids.
Differential modulation by Akkermansia muciniphila and Faecalibacterium prausnitzii of host peripheral lipid metabolism and histone acetylation in mouse gut organoids.
mBio (2014-08-15)
Sabina Lukovac, Clara Belzer, Linette Pellis, Bart J Keijser, Willem M de Vos, Roy C Montijn, Guus Roeselers
ABSTRACT
The gut microbiota is essential for numerous aspects of human health. However, the underlying mechanisms of many host-microbiota interactions remain unclear. The aim of this study was to characterize effects of the microbiota on host epithelium using a novel ex vivo model based on mouse ileal organoids. We have explored the transcriptional response of organoids upon exposure to short-chain fatty acids (SCFAs) and products generated by two abundant microbiota constituents, Akkermansia muciniphila and Faecalibacterium prausnitzii. We observed that A. muciniphila metabolites affect various transcription factors and genes involved in cellular lipid metabolism and growth, supporting previous in vivo findings. Contrastingly, F. prausnitzii products exerted only weak effects on host transcription. Additionally, A. muciniphila and its metabolite propionate modulated expression of Fiaf, Gpr43, histone deacetylases (HDACs), and peroxisome proliferator-activated receptor gamma (Pparγ), important regulators of transcription factor regulation, cell cycle control, lipolysis, and satiety. This work illustrates that specific bacteria and their metabolites differentially modulate epithelial transcription in mouse organoids. We demonstrate that intestinal organoids provide a novel and powerful ex vivo model for host-microbiome interaction studies. We investigated the influence of the gut microbiota and microbially produced short-chain fatty acids (SCFAs) on gut functioning. Many commensal bacteria in the gut produce SCFAs, particularly butyrate, acetate, and propionate, which have been demonstrated to reduce the risk of gastrointestinal disorders. Organoids-small crypt-villus structures grown from ileal intestinal stem cells-were exposed to SCFAs and two specific gut bacteria. Akkermansia muciniphila, found in the intestinal mucus, was recently shown to have a favorable effect on the disrupted metabolism associated with obesity. Faecalibacterium prausnitzii is a commensal gut bacterium, the absence of which may be associated with Crohn's disease. We showed that in our model, A. muciniphila induces stronger effects on the host than F. prausnitzii. We observed that A. muciniphila and propionate affect the expression of genes involved in host lipid metabolism and epigenetic activation or silencing of gene expression. We demonstrated that organoids provide a powerful tool for host-microbe interaction studies.
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FG
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
USP
Sodium acetate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium acetate, anhydrous, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium acetate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium acetate, Vetec, reagent grade, 98%
Sigma-Aldrich
Sodium acetate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium acetate, puriss. p.a., ACS reagent, reag. Ph. Eur., anhydrous
Sigma-Aldrich
Sodium acetate, anhydrous, ≥99%
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Sodium acetate solution, BioUltra, ~3 M in H2O
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%