Merck
  • RETRA exerts anticancer activity in Ewing's sarcoma cells independent of their TP53 status.
RETRA exerts anticancer activity in Ewing's sarcoma cells independent of their TP53 status.
European journal of cancer (Oxford, England : 1990) (2015-03-25)
Jürgen Sonnemann, Désirée Grauel, Lisa Blümel, Julia Hentschel, Christian Marx, Annelie Blumrich, Katharina Focke, Sabine Becker, Susan Wittig, Sandra Schinkel, Oliver H Krämer, James F Beck
ABSTRACT
Mutant p53 can exert oncogenic activity by inhibitory interaction with p73. The small-molecule RETRA has been described to disrupt this interaction and to suppress carcinoma cells (Kravchenko et al., 2008). RETRA's anticancer activity was restricted to tumour cells bearing mutant p53; it was not active in p53 negative and in p53 wild-type cells. Here, we explored the responsiveness of Ewing's sarcoma (ES) cells with mutant p53 to RETRA. For comparison, we also tested RETRA in p53 null and in p53 wild-type ES cells. We found RETRA to be effective in the three mutant p53 ES cell lines investigated. Strikingly, however, RETRA was similarly effective in the p53-deficient and in the two p53 wild-type ES cell lines examined. RETRA elicited apoptosis, as assessed by flow cytometric analyses of mitochondrial depolarisation and DNA fragmentation, caspase 3/7 activity assay and PARP-1 cleavage immunodetection, and G2/M cell cycle arrest completely independent of the cellular TP53 status. In contrast, various p53-deficient and -proficient carcinoma, osteosarcoma and leukaemia cells were unresponsive to RETRA. RETRA also induced gene expression of p53 target genes PUMA and p21 in ES cells irrespective of their TP53 status. These in vitro findings provide a rationale for an in vivo exploration of RETRA's potential as an effective therapeutic approach for patients with ES.
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Formamide, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Nutlin-3, ≥98% (HPLC), powder
SAFC
Sodium chloride solution, 5 M
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Formamide, BioReagent, ≥99.5% (GC), for molecular biology
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Formamide, puriss. p.a., ACS reagent, ≥99.5% (GC/T)
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Formamide, ACS reagent, ≥99.5%
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Glycerol solution, puriss., meets analytical specification of Ph. Eur., BP, 84-88%