Merck
  • Improved analysis of vitamin D metabolites in plasma using liquid chromatography tandem mass spectrometry, and its application to cardiovascular research.
Improved analysis of vitamin D metabolites in plasma using liquid chromatography tandem mass spectrometry, and its application to cardiovascular research.
Biomedical chromatography : BMC (2014-05-28)
Jatinderpal K Sandhu, Janica Auluck, Leong L Ng, Donald J L Jones
ABSTRACT
The accurate and specific measurement of vitamin D is increasingly important for determining the role of vitamin D in the pathogenesis of disease. Liquid chromatography coupled to tandem mass spectrometry (LC/MS/MS) has increasingly become the analytical modality of choice for the analysis of vitamin D. There are many advantages to using LC/MS/MS, such as high specificity and sensitivity to help distinguish the isomers of vitamin D. This rapid method, modified from a Waters Corporation application note, consists of minimal sample manipulation using liquid-liquid extraction and incorporates an internal standard. The supernatant is dried down and injected onto an ultra-high-performance liquid chromatography/electrospray ionization tandem mass spectrometer. The total analysis time is 10 min per injection, enabling high throughput of samples. This method also incorporates two commercial quality control standards to provide a robust system with acceptable coefficient of variation. The analysis of control and heart failure plasma samples showed significant differences in the levels of vitamin D3 between these two groups; however, in the control group, there were individuals who were vitamin D deficient. Overall, the vitamin D3 levels were higher in control samples than in heart failure individuals. As expected, vitamin D2 levels were not observed in many of the samples analysed. This modified method is quick and incorporates an internal standard to allow for any loss in the extraction procedure. The method also includes quality control samples to enable assay standardization. The assay involves inexpensive pre-sample clean-up, aiding high throughput, which is important in many laboratories.
Product Description
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Supelco
Methanol, analytical standard
Sigma-Aldrich
25-Hydroxyvitamin D3 monohydrate, ≥99.0% (HPLC)
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
25-Hydroxyvitamin D2, ≥95.0% (HPLC)
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Supelco
2-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
25-Hydroxyvitamin D2 solution, 100 μg/mL in ethanol, 98% (CP)
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
25-Hydroxyvitamin D2 solution, 50 μg/mL in ethanol, 98% (CP)
Sigma-Aldrich
25-Hydroxyvitamin D2 solution, 5 μg/mL in ethanol, 98% (CP)
Supelco
2-Propanol, analytical standard
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
25-Hydroxycholecalciferol, ≥98% (HPLC)
Sigma-Aldrich
2-Propanol, for molecular biology, BioReagent, ≥99.5%