Merck
  • Dual role of photosensitizer and carrier material of fullerene in micelles for chemo-photodynamic therapy of cancer.
Dual role of photosensitizer and carrier material of fullerene in micelles for chemo-photodynamic therapy of cancer.
Journal of pharmaceutical sciences (2014-09-02)
Xinhong Guo, Rui Ding, Ying Zhang, Ling Ye, Xin Liu, Caiping Chen, Zhenzhong Zhang, Yun Zhang
ABSTRACT
Derivatives of fullerene (C60) as photosensitizers have rarely been studied as delivery carrier materials. The focus of this study was to explore the potential advantages of diadduct malonic acid-fullerene (DMA-C60) as delivery carrier materials and combination of chemo-phototherapy of some tumors. In this study, DMA-C60 and docetaxel (DTX) were coentrapped in micelles (MCs) (DMA-C60/DTX-MC). The addition of DMA-C60 could obviously improve static stability and decrease critical MC concentration of DTX-MC without hemolysis. The sustained release of DTX and DMA-C60 could be achieved, following Higuichi and first-order model, respectively. DMA-C60 could still produce reactive oxygen species efficiently in HeLa cells after encapsulation in MC. The addition of DMA-C60 under irradiation caused DTX-MC more stronger cytotoxicity, cell cycle changes, and more early apoptotic cells in vitro. More importantly, after intravenous injection, the addition of DMA-C60 in DTX-MC could result in 2.25-fold and 4.57-fold longer mean residence time compared with DTX-MC and Duopafei(®) , increase drug intratumoral distribution and decrease drug distribution in heart and kidney, and enhance antitumor effect under irradiation without body weight loss. These results suggested tremendous promise of DMA-C60 as carrier materials of MC and significant advantages in combination of chemo-phototherapy of some tumors. © 2014 Wiley Periodicals, Inc. and the American Pharmacists Association J Pharm Sci 103:3225-3234, 2014.
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Supelco
Methanol, analytical standard
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Sulforhodamine B, Dye content 75 %
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Sulforhodamine B sodium salt, powder, BioReagent, suitable for cell culture