Merck
  • Discovery and characterization of auxiliary proteins encoded by type 3 simian T-cell lymphotropic viruses.
Discovery and characterization of auxiliary proteins encoded by type 3 simian T-cell lymphotropic viruses.
Journal of virology (2014-10-31)
Jocelyn Turpin, Chloé Journo, Nga Ling Ko, Flore Sinet, Alexandre Carpentier, Amandine Galioot, Dustin Edwards, Anne-Mieke Vandamme, Louis Gazzolo, Madeleine Duc Dodon, Antoine Gessain, Fatah Kashanchi, Ivan Balansard, Romain Lacoste, Renaud Mahieux
ABSTRACT
Human T-cell lymphotropic virus type 1 (HTLV-1) and HTLV-2 encode auxiliary proteins that play important roles in viral replication, viral latency, and immune escape. The presence of auxiliary protein-encoding open reading frames (ORFs) in HTLV-3, the latest HTLV to be discovered, is unknown. Simian T-cell lymphotropic virus type 3 (STLV-3) is almost identical to HTLV-3. Given the lack of HTLV-3-infected cell lines, we took advantage of STLV-3-infected cells and of an STLV-3 molecular clone to search for the presence of auxiliary transcripts. Using reverse transcriptase PCR (RT-PCR), we first uncovered the presence of three unknown viral mRNAs encoding putative proteins of 5, 8, and 9 kDa and confirmed the presence of the previously reported RorfII transcript. The existence of these viral mRNAs was confirmed by using splice site-specific RT-PCR with ex vivo samples. We showed that p5 is distributed throughout the cell and does not colocalize with a specific organelle. The p9 localization is similar to that of HTLV-1 p12 and induced a strong decrease in the calreticulin signal, similarly to HTLV-1 p12. Although p8, RorfII, and Rex-3 share an N-terminal sequence that is predicted to contain a nucleolar localization signal (NoLS), only p8 is found in the nucleolus. The p8 location in the nucleolus is linked to a bipartite NoLS. p8 and, to a lesser extent, p9 repressed viral expression but did not alter Rex-3-dependent mRNA export. Using a transformation assay, we finally showed that none of the STLV-3 auxiliary proteins had the ability to induce colony formation, while both Tax-3 and antisense protein of HTLV-3 (APH-3) promoted cellular transformation. Altogether, these results complete the characterization of the newly described primate T-lymphotropic virus type 3 (PTLV-3). Together with their simian counterparts, HTLVs form the primate T-lymphotropic viruses. HTLVs arose from interspecies transmission between nonhuman primates and humans. HTLV-1 and HTLV-2 encode auxiliary proteins that play important roles in viral replication, viral latency, and immune escape. The presence of ORFs encoding auxiliary proteins in HTLV-3 or STLV-3 genomes was unknown. Using in silico analyses, ex vivo samples, or in vitro experiments, we have uncovered the presence of 3 previously unknown viral mRNAs encoding putative proteins and confirmed the presence of a previously reported viral transcript. We characterized the intracellular localization of the four proteins. We showed that two of these proteins repress viral expression but that none of them have the ability to induce colony formation. However, both Tax and the antisense protein APH-3 promote cell transformation. Our results allowed us to characterize 4 new retroviral proteins for the first time.
Product Description
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Alanine, European Pharmacopoeia (EP) Reference Standard
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Sodium Fluoride Solution
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Sodium fluoride 0.5 M solution
Supelco
Fluoride ion solution for ISE, 0.1 M F-, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
DL-Alanine, ≥99%, FCC, FG
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Sigma-Aldrich
Sodium fluoride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Anti-HA antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Sodium fluoride, ReagentPlus®, ≥99%
Sigma-Aldrich
DL-Alanine, ≥99% (HPLC)
Supelco
Sodium Fluoride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium fluoride, puriss., meets analytical specification of Ph. Eur., BP, USP, 98.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium fluoride, ACS reagent, ≥99%