Merck
  • Disequilibrium of M1 and M2 macrophages correlates with the development of experimental inflammatory bowel diseases.
Disequilibrium of M1 and M2 macrophages correlates with the development of experimental inflammatory bowel diseases.
Immunological investigations (2014-06-13)
Wei Zhu, Jianbo Yu, Ying Nie, XueKui Shi, Yang Liu, Fujuan Li, Xiao-li Zhang
ABSTRACT
Ulcerative colitis, a major inflammatory bowel disease, is an idiopathic inflammatory disorder of the colonic mucosa, accompanied by an aberrant immune reaction to intestinal microflora. Macrophages are central mediators of intestinal immune homeostasis and inflammation. The relationship between macrophages and the pathogenesis of colitis is poorly understood. We aimed to characterize the changing populations and roles of M1/M2 macrophages in colitis. We demonstrated that M1 macrophages increased and M2 macrophages decreased in colitis, accompanied by Interleukin (IL)-23 and Tumor necrosis factor-α induction and IL-10 suppression. Transfer of M2 macrophages reduced dextran sodium sulfate-induced colitis by inducing IL-10 production and promoting regulatory T-cell generation. In vivo neutralization of IL-10 partially reduced the effects of M2 transfer. These findings suggest that macrophages play a critical role in colitis; specifically, disequilibrium of macrophage subsets promotes colitis development. A shift from the M1 to M2 phenotype reduces colitis by inducing IL-10; thus, mobilization of M2 macrophages could be a novel approach to colitis therapy.
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium sulfate, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Sodium sulfate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Sodium sulfate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium sulfate, puriss., meets analytical specification of Ph. Eur., BP, USP, anhydrous, 99.0-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, granular
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, powder
Sigma-Aldrich
Sodium sulfate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Rat IL-10 ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Rat IL-10 ELISA Kit, for cell and tissue lysates
Sigma-Aldrich
Human IL-10 R β ELISA Kit, for serum, plasma, cell culture supernatant and urine
Sigma-Aldrich
Mouse IL-10 ELISA Kit, for serum, plasma and cell culture supernatant