MilliporeSigma
  • The ether lipid precursor hexadecylglycerol protects against Shiga toxins.
The ether lipid precursor hexadecylglycerol protects against Shiga toxins.
Cellular and molecular life sciences : CMLS (2014-04-18)
Jonas Bergan, Tore Skotland, Anne Berit Dyve Lingelem, Roger Simm, Bjørn Spilsberg, Toril Lindbäck, Tuulia Sylvänne, Helena Simolin, Kim Ekroos, Kirsten Sandvig
ABSTRACT
Shiga toxin-producing Escherichia coli bacteria cause hemorrhagic colitis and hemolytic uremic syndrome in humans. Currently, only supportive treatment is available for diagnosed patients. We show here that 24-h pretreatment with an ether lipid precursor, the alkylglycerol sn-1-O-hexadecylglycerol (HG), protects HEp-2 cells against Shiga toxin and Shiga toxin 2. Also the endothelial cell lines HMEC-1 and HBMEC are protected against Shiga toxins after HG pretreatment. In contrast, the corresponding acylglycerol, DL-α-palmitin, has no effect on Shiga toxicity. Although HG treatment provides a strong protection (~30 times higher IC₅₀) against Shiga toxin, only a moderate reduction in toxin binding was observed, suggesting that retrograde transport of the toxin from the plasma membrane to the cytosol is perturbed. Furthermore, endocytosis of Shiga toxin and retrograde sorting from endosomes to the Golgi apparatus remain intact, but transport from the Golgi to the endoplasmic reticulum is inhibited by HG treatment. As previously described, HG reduces the total level of all quantified glycosphingolipids to 50-70% of control, including the Shiga toxin receptor globotriaosylceramide (Gb3), in HEp-2 cells. In accordance with this, we find that interfering with Gb3 biosynthesis by siRNA-mediated knockdown of Gb3 synthase for 24 h causes a similar cytotoxic protection and only a moderate reduction in toxin binding (to 70% of control cells). Alkylglycerols, including HG, have been administered to humans for investigation of therapeutic roles in disorders where ether lipid biosynthesis is deficient, as well as in cancer therapy. Further studies may reveal if HG can also have a therapeutic potential in Shiga toxin-producing E. coli infections.
Product Description
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
SKU
Pack Size
Availability
Price
Quantity
Hydrocortisone, European Pharmacopoeia (EP) Reference Standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ruthenium on carbon, extent of labeling: 5 wt. % loading
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ruthenium black
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, ≥99% (HPLC), powder
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, BioUltra, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
USP
Hydrocortisone, United States Pharmacopeia (USP) Reference Standard
SKU
Pack Size
Availability
Price
Quantity
Hydrocortisone, British Pharmacopoeia (BP) Assay Standard
SKU
Pack Size
Availability
Price
Quantity
Leucine, European Pharmacopoeia (EP) Reference Standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
MISSION® esiRNA, targeting mouse A4galt
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
DAPI, for nucleic acid staining
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
DL-Leucine, ≥99% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate tris salt, ≥97% (HPLC), powder
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ruthenium, powder, −200 mesh, 99.9% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
MISSION® esiRNA, targeting human A4GALT
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate, ≥98.5% (HPLC), powder
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ruthenium, powder, 99.99% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Hydrocortisone for peak identification, European Pharmacopoeia (EP) Reference Standard
SKU
Pack Size
Availability
Price
Quantity
Ruthenium, Ruthenium, foil, 6x6mm, thickness 1.0mm, 99.9%
SKU
Pack Size
Availability
Price
Quantity
Ruthenium, Ruthenium, microfoil, disks, 10mm, thinness 0.025μm, specific density 30.5μg/cm2, permanent mylar 3.5μm support, 99.9%
SKU
Pack Size
Availability
Price
Quantity
Ruthenium, Ruthenium, microfoil, disks, 10mm, thinness 0.1μm, specific density 122μg/cm2, permanent mylar 3.5μm support, 99.9%
SKU
Pack Size
Availability
Price
Quantity
Ruthenium, Ruthenium, pellets, 2.5g, max. size 10mm, 99.9%
SKU
Pack Size
Availability
Price
Quantity
Ruthenium, Ruthenium, foil, 25x25mm, thickness 1.0mm, 99.9%
SKU
Pack Size
Availability
Price
Quantity
Ruthenium, Ruthenium, pellets, 5g, max. size 10mm, 99.9%
SKU
Pack Size
Availability
Price
Quantity
Ruthenium, Ruthenium, rod, 12.7mm, diameter 12.7mm, 99.9%
SKU
Pack Size
Availability
Price
Quantity
Ruthenium, Ruthenium, bar, 25mm x 2mm x 2mm, 99.9%