Merck
Phosphatidylethanolamine positively regulates autophagy and longevity.
Cell death and differentiation (2015-01-13)
P Rockenfeller, M Koska, F Pietrocola, N Minois, O Knittelfelder, V Sica, J Franz, D Carmona-Gutierrez, G Kroemer, F Madeo
ABSTRACT
Autophagy is a cellular recycling program that retards ageing by efficiently eliminating damaged and potentially harmful organelles and intracellular protein aggregates. Here, we show that the abundance of phosphatidylethanolamine (PE) positively regulates autophagy. Reduction of intracellular PE levels by knocking out either of the two yeast phosphatidylserine decarboxylases (PSD) accelerated chronological ageing-associated production of reactive oxygen species and death. Conversely, the artificial increase of intracellular PE levels, by provision of its precursor ethanolamine or by overexpression of the PE-generating enzyme Psd1, significantly increased autophagic flux, both in yeast and in mammalian cell culture. Importantly administration of ethanolamine was sufficient to extend the lifespan of yeast (Saccharomyces cerevisiae), mammalian cells (U2OS, H4) and flies (Drosophila melanogaster). We thus postulate that the availability of PE may constitute a bottleneck for functional autophagy and that organismal life or healthspan could be positively influenced by the consumption of ethanolamine-rich food.
Sigma-Aldrich
Rapamycin, Ready Made Solution, 2.5 mg/mL in DMSO (2.74 mM), from Streptomyces hygroscopicus
Supelco
Aucubin, analytical standard
Sigma-Aldrich
Ethanolamine, ACS reagent, ≥99.0%
Sigma-Aldrich
Ethanolamine, liquid, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Ethanolamine, ≥98%
Aucubin, primary reference standard
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, purum p.a., ≥35% (RT)
Avanti
Egg Trans PE, L-α-phosphatidylethanolamine, transphosphatidylated (Egg, Chicken), chloroform
Avanti
Egg Trans PE, L-α-phosphatidylethanolamine, transphosphatidylated (Egg, Chicken), powder
Sigma-Aldrich
Ethanolamine, puriss. p.a., ACS reagent, ≥99.0% (GC/NT)
Supelco
Ethanolamine, analytical standard
Sigma-Aldrich
Ethanolamine, purified by redistillation, ≥99.5%
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Ethanolamine, ≥99%
Sigma-Aldrich
Nitrogen, ≥99.998%
Sigma-Aldrich
N-[Tris(hydroxymethyl)methyl]acrylamide, contains ≤7% KCl, 93%
Sigma-Aldrich
Ethanolamine hydrochloride, ≥99.0%
Supelco
Rapamycin, VETRANAL®, analytical standard
Trolamine impurity A, European Pharmacopoeia (EP) Reference Standard
Roche
Anti-GFP, from mouse IgG1κ (clones 7.1 and 13.1)
Sigma-Aldrich
Ethanolamine hydrochloride, anhydrous, free-flowing, Redi-Dri, ≥99.0%