Merck
  • Pyrrolidine dithiocarbamate inhibits UVB-induced skin inflammation and oxidative stress in hairless mice and exhibits antioxidant activity in vitro.
Pyrrolidine dithiocarbamate inhibits UVB-induced skin inflammation and oxidative stress in hairless mice and exhibits antioxidant activity in vitro.
Journal of photochemistry and photobiology. B, Biology (2014-06-14)
Ana L M Ivan, Marcela Z Campanini, Renata M Martinez, Vitor S Ferreira, Vinicius S Steffen, Fabiana T M C Vicentini, Fernanda M P Vilela, Frederico S Martins, Ana C Zarpelon, Thiago M Cunha, Maria J V Fonseca, Marcela M Baracat, Sandra R Georgetti, Waldiceu A Verri, Rúbia Casagrande
ABSTRACT
Ultraviolet B (UVB) irradiation may cause oxidative stress- and inflammation-dependent skin cancer and premature aging. Pyrrolidine dithiocarbamate (PDTC) is an antioxidant and inhibits nuclear factor-κB (NF-κB) activation. In the present study, the mechanisms of PDTC were investigated in cell free oxidant/antioxidant assays, in vivo UVB irradiation in hairless mice and UVB-induced NFκB activation in keratinocytes. PDTC presented the ability to scavenge 2,2'-azinobis-(3-ethyl benzothiazoline-6-sulfonic acid) radical (ABTS), 2,2-diphenyl-1-picryl-hydrazyl radical (DPPH) and hydroxyl radical (OH); and also efficiently inhibited iron-dependent and -independent lipid peroxidation as well as chelated iron. In vivo, PDTC treatment significantly decreased UVB-induced skin edema, myeloperoxidase (MPO) activity, production of the proinflammatory cytokine interleukin-1β (IL-1β), matrix metalloproteinase-9 (MMP-9), increase of reduced glutathione (GSH) levels and antioxidant capacity of the skin tested by the ferric reducing antioxidant power (FRAP) and ABTS assays. PDTC also reduced UVB-induced IκB degradation in keratinocytes. These results demonstrate that PDTC presents antioxidant and anti-inflammatory effects in vitro, which line up well with the PDTC inhibition of UVB irradiation-induced skin inflammation and oxidative stress in mice. These data suggest that treatment with PDTC may be a promising approach to reduce UVB irradiation-induced skin damages and merits further pre-clinical and clinical studies.
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Supelco
Linoleic acid, analytical standard
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ReagentPlus®, 99%
Sigma-Aldrich
Linoleic acid, liquid, BioReagent, suitable for cell culture
Sigma-Aldrich
Linoleic acid, ≥99%
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ≥98%, BioReagent, suitable for determination of sulfhydryl groups
Sigma-Aldrich
o-Dianisidine dihydrochloride, ≥95%
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
2-Thiobarbituric acid, ≥98%
Sigma-Aldrich
o-Dianisidine dihydrochloride, Suitable for use in glucose determination
Sigma-Aldrich
o-Dianisidine dihydrochloride, tablet, 10 mg substrate per tablet
Sigma-Aldrich
Linoleic acid, technical, 58-74% (GC)
Isoflurane, European Pharmacopoeia (EP) Reference Standard
Glutathione, European Pharmacopoeia (EP) Reference Standard
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Cetrimonium bromide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), Vetec, reagent grade, 98%
Supelco
Hexadecyltrimethylammonium bromide, analytical standard
Sigma-Aldrich
1,10-Phenanthroline monohydrate, ACS reagent, puriss. p.a., ≥99.5% (calc. to the dried substance), for redox titration
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥96.0% (AT)
Supelco
Hexadecyltrimethylammonium bromide, suitable for ion pair chromatography, LiChropur
Sigma-Aldrich
N-Ethylmaleimide, purum p.a., ≥99.0% (HPLC)
Sigma-Aldrich
N-Ethylmaleimide, BioUltra, ≥99.0% (HPLC)
Supelco
Bathophenanthroline, for spectrophotometric det. of Fe in serum, ≥99.0%
Supelco
o-Xylene, analytical standard
Sigma-Aldrich
1,1-Diphenyl-2-picrylhydrazine, 97%
Sigma-Aldrich
1,10-Phenanthroline monohydrate, ACS reagent, 99%
Sigma-Aldrich
N-Ethylmaleimide, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
N-Ethylmaleimide, crystalline, ≥98% (HPLC)
Sigma-Aldrich
2,2-Diphenyl-1-picrylhydrazyl